Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  60  67 
1171 - 1200 of 2000 products displayed
  • ZO-1 Antibody (Monoclonal, ZO1-1A12)
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated See Additional Formats Alexa Fluor 488 Alexa Fluor 594 FITC
  • Catalog # 339100
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
339100 311.00
Check your price
  • Tau Antibody (Monoclonal, TAU-5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # AHB0042
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHB0042 311.00
Check your price
  • Human TNF alpha Recombinant Protein
  • Host Yeast
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RTNFAI
  • Size 20 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RTNFAI 310.00
Check your price
  • Human IFN alpha 2a Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 111012
  • Size 20 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Interferon alpha-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
111012 310.00
Check your price
  • TRPV5 Antibody
  • Host Rabbit
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # OST00148W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:3000), Western Blot (1:300-1:3000)
Alternate names:
Transient receptor potential cation channel subfamily V member 5, TrpV5, osm-9-like TRP channel 3, OTRPC3, Epithelial calcium channel 1, ECaC1, Calcium transporter 2, CaT2, Ecac,CAT2, ECAC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OST00148W 310.00
Check your price
  • PSD-95 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 516900
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (0.5-1.0 ug/ml), Immunoprecipitation (5 ug), Western Blot (1.0 ug/ml)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
516900 310.00
Check your price
  • Occludin Antibody (Monoclonal, OC-3F10)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated See Additional Formats Alexa Fluor 488 Alexa Fluor 594 FITC HRP
  • Catalog # 331500
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunocytochemistry (2-3 µg/ml), Immunofluorescence (2-3 ug/ml), Western Blot (0.1-1.0 ug/ml)
Alternate names:
OCLN
Catalog # Size Price (AUD) Your Price (AUD) Qty
331500 310.00
Check your price
  • alpha Synuclein Antibody (Monoclonal, Syn 211)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 328100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
328100 310.00
Check your price
  • CD3/CD20 Antibody (Cocktail, CD3-SP7; CD20-L26)
  • Host Mixed
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514433
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Neat), Western Blot (Neat-1:100)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; Membrane-spanning 4-domains subfamily A member 1; Leukocyte surface antigen Leu-16; Bp35; B-lymphocyte surface antigen B1; Lymphocyte antigen 44; B-cell differentiation antigen Ly-44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514433 309.00
Check your price
  • Lysozyme Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126309
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
1,4-beta-N-acetylmuramidase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126309 309.00
Check your price
  • HSV Type 1 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA129210
  • Size 6 mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129210 309.00
Check your price
  • Vimentin Antibody (Monoclonal, SP20)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514564
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
VIM, FLJ36605
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514564 309.00
Check your price
  • VEGF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516754
  • Size 500 µL
Applications
Immunocytochemistry (1:40-1:200), Immunofluorescence (1:40-1:200), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (3 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Vascular permeability factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516754 309.00
Check your price
  • TFF1 Antibody (Monoclonal, pS2.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511507
  • Size 500 µL
Applications
Immunohistochemistry (Assay Dependent)
Alternate names:
Protein pS2; Polypeptide P1.A; PNR-2; Breast cancer estrogen-inducible protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511507 309.00
Check your price
  • TAG72 Antibody (Monoclonal, B72.3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512065
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512065 309.00
Check your price
  • Plasma Cell Antibody (Monoclonal, LIV3G11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA513532
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513532 309.00
Check your price
  • p53 Antibody (Monoclonal, DO-1)
  • Host Mouse
  • Target Species Human, Rhesus monkey
  • Conjugate Unconjugated
  • Catalog # MA512571
  • Size 500 µL
Applications
ChIP assay (1-3 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512571 309.00
Check your price
  • NF1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA146246
  • Size 500 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:200-1:500), Immunohistochemistry (Paraffin) (1:200-1:500), Western Blot (1:500-1:2000)
Alternate names:
Neurofibromatosis-related protein NF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146246 309.00
Check your price
  • Myosin Skeletal Muscle Antibody (Monoclonal, MYSN02 (MY-32))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511748
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:500-1:1000)
Alternate names:
Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain IIx/d; Myosin heavy chain 2x; Myosin heavy chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511748 309.00
Check your price
  • Melan-A Antibody (Monoclonal, A103)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514168
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100-200), Immunoprecipitation (2 µg/ml), Western Blot (1 µg/mL)
Alternate names:
Protein Melan-A; Antigen SK29-AA; Antigen LB39-AA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514168 309.00
Check your price
  • MAP2 Antibody (Monoclonal, AP18)
  • Host Mouse
  • Target Species Avian, Bovine, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA512826
  • Size 500 µL
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
MAP-2,OTTHUMP00000208143
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512826 309.00
Check your price
  • Laminin beta-2/gamma-1 Antibody (Monoclonal, A5)
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514649
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (2 µg/ml), Immunoprecipitation (2 µg/ml)
Alternate names:
S-LAM beta, S-laminin subunit beta, laminin B1s chain, laminin S, laminin subunit beta-2, laminin-11 subunit beta, laminin-14 subunit beta, laminin-15 subunit beta, laminin-3 subunit beta, laminin-4 subunit beta, laminin-7 subunit beta, laminin-9 subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514649 309.00
Check your price
  • Ki-67 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516446
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (10-20 µg/mL)
Alternate names:
antigen KI-67, proliferation-related Ki-67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516446 309.00
Check your price
  • Histone H1 Antibody (Monoclonal, 1415-1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513750
  • Size 500 µL
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:100-1:500)
Alternate names:
TISP64
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513750 309.00
Check your price
  • HGF Antibody (Monoclonal, SBF5)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514160
  • Size 500 µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Western Blot (Assay Dependent)
Alternate names:
Scatter factor; Hepatopoietin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514160 309.00
Check your price
  • Helicobacter pylori Antibody
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA532442
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
H. pylori, Campylobacter pyloridis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532442 309.00
Check your price
  • Cytokeratin 8/18 Antibody (Cocktail, K8.8 + DC10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512281
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512281 309.00
Check your price
  • Cytokeratin 13 Antibody (Monoclonal, 34BetaE12)
  • Host Mouse
  • Target Species Horse, Human, Non-human primate, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA512135
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:1000-1:10,000)
Alternate names:
Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512135 309.00
Check your price
  • COL7A1 Antibody (Monoclonal, LH7.2)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Goat, Human, Non-human primate, Sheep, Pig
  • Conjugate Unconjugated
  • Catalog # MA513432
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunomicroscopy (Assay Dependent)
Alternate names:
Long-chain collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513432 309.00
Check your price
  • Chromogranin A Antibody (Monoclonal, SP12)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA514536
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:5000), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
Pituitary secretory protein I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514536 309.00
Check your price