Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  110  117 
301 - 330 of 3500 products displayed
  • Goat anti-Human IgG / IgA / IgM Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # A18848
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18848 94.00
Check your price
  • ZMYND10 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525732
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein BLu
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525732 349.00
Check your price
  • WNT5A Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515511
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
hWNT5A; WNT5A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515511 349.00
Check your price
  • SEMA5A Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547791
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Semaphorin-F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547791 349.00
Check your price
  • SEMA5A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA530884
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Semaphorin-F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530884 349.00
Check your price
  • PRG2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525636
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Proteoglycan 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525636 349.00
Check your price
  • NOXA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA118332
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Protein Noxa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118332 349.00
Check your price
  • MYO1C Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513384
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Myosin I beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513384 349.00
Check your price
  • Lactoferrin Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA140280
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
GIG12, HLF2, LF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA140280 550.00
Check your price
  • Phospho-JAK2 (Tyr1007) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536658
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Janus kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536658 349.00
Check your price
  • IL-31 Polyclonal Antibody for Western Blot, IF, ICC, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547405
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Neutralization (1 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
interleukin-31
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547405 349.00
Check your price
  • IL1RA Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547661
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Neutralization (1-5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
IL1 inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547661 349.00
Check your price
  • IL-17F Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516228
  • Size 100ug
Applications
Western Blot (0.2-2 µg/ml)
Alternate names:
Cytokine ML-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516228 349.00
Check your price
  • ICAM-3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529817
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
ICAM-R; CDw50
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529817 349.00
Check your price
  • Phospho-Histone H2A.X (Ser139) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFPA0025
  • Size 100µL
Applications
Western Blot (1:5000)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0025 309.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Cat, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA316727
  • Size 50µL
Applications
Immunocytochemistry (1:1000-1:5000), Immunofluorescence (1:1000-1:5000), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000-1:5000), Western Blot (1:50,000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316727 375.00
Check your price
  • GFAP Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Bovine, Chicken, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA110019
  • Size 100µL
Applications
Immunocytochemistry (1:1000-1:5000), Immunofluorescence (1:1000-1:5000), Immunohistochemistry (1:1000-1:5000), Western Blot (1:10,000-1:100,000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110019 309.00
Check your price
  • GFAP Monoclonal Antibody for Western Blot, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106701
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (1:25-1:100), Immunohistochemistry (Paraffin) (1:25-1:100), Western Blot (1:100-1:1000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106701 349.00
Check your price
  • EMB Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531922
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
embigin homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531922 349.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516429
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (10 µg/ml), Western Blot (5-10 µg/mL)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516429 349.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516762
  • Size 500µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:250-1:500), Immunoprecipitation (Assay Dependent), Western Blot (2 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516762 349.00
Check your price
  • Phospho-beta Catenin (Ser33) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537543
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537543 309.00
Check your price
  • Phospho-beta Catenin (Ser33, Ser37) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Chicken, Human, Marsupial, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523354
  • Size 20ug
Applications
Western Blot (0.25-1 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523354 349.00
Check your price
  • GM-CSF Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MM500C
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM500C 349.00
Check your price
  • Phospho-CREB (Ser129, Ser133) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA124754
  • Size 50µL
Applications
Western Blot (1:1000)
Alternate names:
CREB, MGC9284
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124754 349.00
Check your price
  • CD37 Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515492
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000)
Alternate names:
Tetraspanin-26
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515492 349.00
Check your price
  • ASL Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522052
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Arginosuccinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522052 349.00
Check your price
  • beta-Arrestin 2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1732
  • Size 100µg
Applications
Western Blot (1:2000)
Alternate names:
Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1732 349.00
Check your price
  • beta Actin Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate HRP
  • Catalog # MA515739HRP
  • Size 50µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739HRP 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA5157391MG
  • Size 1mg
Applications
ELISA (Assay dependent), Flow Cytometry (1-2 ug/test), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5157391MG 950.00
Check your price