Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  110  117 
361 - 390 of 3500 products displayed
  • Trans-Golgi Network 38 Polyclonal Antibody for Western Blot, IF, IHC (F) and IM
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184496
  • Size 100µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1/100-1/200), Immunomicroscopy (Assay Dependent), Western Blot (1/1000)
Alternate names:
TGN38B; TGN38A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184496 550.00
Check your price
  • SERPINB7 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531891
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
TP55; Megsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531891 349.00
Check your price
  • RAD51 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514419
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514419 349.00
Check your price
  • NPY2R Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526477
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
NPY-Y2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526477 349.00
Check your price
  • NPY2R Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141576
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
NPY-Y2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141576 375.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511518
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511518 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520636
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520636 349.00
Check your price
  • KLHL22 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515699
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
KELCHL; KLHL22
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515699 349.00
Check your price
  • ITGB6 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547588
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547588 349.00
Check your price
  • ITGB5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527759
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
integrin beta-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527759 349.00
Check your price
  • IL-20 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547067
  • Size 100µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytokine Zcyto10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547067 349.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MP700
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; interferon gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MP700 349.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MP701
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; interferon gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MP701 349.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MM701
  • Size 500ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; gamma interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM701 275.00
Check your price
  • Phospho-HDAC5 (Ser498) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536647
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536647 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530928
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530928 349.00
Check your price
  • CD64 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # CD6404
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6404 263.00
Check your price
  • CD64 Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # CD6406
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6406 321.00
Check your price
  • CrkL Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515867
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Western Blot (1/500-1/2000)
Alternate names:
crk-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515867 349.00
Check your price
  • COMP Monoclonal Antibody for Western Blot, IHC (F), IHC (P), IP and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120221
  • Size 1mL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Thrombospondin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120221 349.00
Check your price
  • CDH4 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511433
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Retinal cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511433 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535089
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535089 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519583
  • Size 100µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunoprecipitation (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519583 349.00
Check your price
  • Septin-8 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531356
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
2-Sep, septin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531356 349.00
Check your price
  • Hepatitis E Virus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA121400
  • Size 250µg
Applications
ELISA (Assay dependent)
Alternate names:
HepE
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121400 399.00
Check your price
  • Interferon gamma (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75631
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75631 2,600.00
Check your price
  • Fluorescein/Oregon Green Polyclonal Antibody for ICC, IHC and Flow
  • Host Rabbit
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # A6413
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A6413 399.00
Check your price
  • Mouse anti-Human IgG, Fd Secondary Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # SA510191
  • Size 50ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510191 170.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 626520
  • Size 1mL
Applications
ELISA (1:2,000-1:4,000), Immunohistochemistry (1:2,000-1:4,000), Western Blot (1:2,000-1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
626520 149.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Biotin-XX conjugate for IF and ICC
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin-XX
  • Catalog # B2770
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
B2770 139.00
Check your price