Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  40  50  60  70  80  90  100  110  117 
481 - 510 of 3500 products displayed
  • K-cadherin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547373
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Kidney cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547373 349.00
Check your price
  • ADAM12 Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524285
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml)
Alternate names:
Meltrin-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524285 275.00
Check your price
  • Gentamycin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA110726
  • Size 500µg
Applications
ELISA (Assay dependent)
Alternate names:
Gentamycin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110726 349.00
Check your price
  • Diphtheria Toxin A Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA146399
  • Size 100µg
Applications
ELISA (1:100-1:2000)
Alternate names:
DT
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146399 309.00
Check your price
  • Canine Parvovirus Monoclonal Antibody for IHC (P) and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA191114
  • Size 250µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
CPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191114 499.00
Check your price
  • Yellow Fever Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17398
  • Size 100µg
Applications
ELISA (1:20-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17398 349.00
Check your price
  • FITC Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical, Tag
  • Conjugate Unconjugated
  • Catalog # MIF2902
  • Size 1mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIF2902 600.00
Check your price
  • Fluorescein/Oregon Green Polyclonal Antibody for ICC, IHC and Flow
  • Host Rabbit
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # A889
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A889 338.00
Check your price
  • M13 Phage coat protein Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # MA134468
  • Size 50µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 µg for 10^6 cells), Immunohistochemistry (Assay dependent), Western Blot (1-5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134468 375.00
Check your price
  • Dehydroepiandrosterone Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA174191
  • Size 200µg
Applications
ELISA (1:1000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174191 325.00
Check your price
  • Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18862
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18862 263.00
Check your price
  • Goat anti-Human Kappa Light Chain Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18856
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18856 220.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Texas Red®-X conjugate for IF
  • Host Goat
  • Target Species Mouse
  • Conjugate Texas Red®-X
  • Catalog # T6390
  • Size 500µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
T6390 138.00
Check your price
  • Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18859
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18859 229.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Rhodamine Red-X conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Rhodamine Red-X
  • Catalog # R6393
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
R6393 137.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC and IHC
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 555
  • Catalog # A21432
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21432 209.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 568
  • Catalog # A11057
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11057 209.00
Check your price
  • Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and IHC
  • Host Rabbit
  • Target Species Goat
  • Conjugate Alexa Fluor® 594
  • Catalog # A11080
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11080 169.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 680
  • Catalog # A21084
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21084 209.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MH10511
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MH10511 147.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517608
  • Size 200ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517608 325.00
Check your price
  • RAD51 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514419
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514419 349.00
Check your price
  • NPY2R Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141576
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
NPY-Y2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141576 375.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511518
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511518 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520636
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520636 349.00
Check your price
  • ITGB6 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547588
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547588 349.00
Check your price
  • IL-20 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547067
  • Size 100µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytokine Zcyto10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547067 349.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MP700
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; interferon gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MP700 349.00
Check your price
  • Phospho-HDAC5 (Ser498) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536647
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536647 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530928
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530928 349.00
Check your price