Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  40  50  60  70  80  90  100  110  117 
511 - 540 of 3500 products displayed
  • M13 Phage coat protein Monoclonal Antibody, Biotin conjugate for Western Blot, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # MA134468
  • Size 50µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 µg for 10^6 cells), Immunohistochemistry (Assay dependent), Western Blot (1-5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134468 375.00
Check your price
  • Dehydroepiandrosterone Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA174191
  • Size 200µg
Applications
ELISA (1:1000-1:10000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174191 325.00
Check your price
  • Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18862
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18862 263.00
Check your price
  • Goat anti-Human Kappa Light Chain Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate AP
  • Catalog # A18856
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18856 220.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Texas Red®-X conjugate for IF
  • Host Goat
  • Target Species Mouse
  • Conjugate Texas Red®-X
  • Catalog # T6390
  • Size 500µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
T6390 138.00
Check your price
  • Goat anti-Human Kappa Light Chain Cross-Adsorbed Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate HRP
  • Catalog # A18859
  • Size 500µg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18859 229.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Rhodamine Red-X conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Rhodamine Red-X
  • Catalog # R6393
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
R6393 137.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC, IHC and Flow
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 568
  • Catalog # A11057
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11057 209.00
Check your price
  • Rabbit anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC and IHC
  • Host Rabbit
  • Target Species Goat
  • Conjugate Alexa Fluor® 594
  • Catalog # A11080
  • Size 500µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11080 169.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and ICC
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 680
  • Catalog # A21084
  • Size 500µL
Applications
Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21084 209.00
Check your price
  • Mouse anti-Human Kappa Light Chain Secondary Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MH10511
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MH10511 147.00
Check your price
  • VAMP1 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517608
  • Size 200ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Synaptobrevin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517608 325.00
Check your price
  • 53BP1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, IP and ChIP
  • Host Rabbit
  • Target Species Fish, Goat, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116565
  • Size 100µL
Applications
Flow Cytometry (1.5 µg/ml), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:400-1:2000), Western Blot (1:5000-1:25,000)
Alternate names:
TP53BP1, p202
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116565 375.00
Check your price
  • RAD51 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514419
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/ml), Western Blot (1:50-1:500)
Alternate names:
RAD51 homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514419 349.00
Check your price
  • NPY2R Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141576
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
NPY-Y2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141576 375.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511518
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511518 349.00
Check your price
  • METTL7A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520636
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (2 µg/ml)
Alternate names:
Protein AAM-B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520636 349.00
Check your price
  • ITGB6 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547588
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
integrin beta 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547588 349.00
Check your price
  • IL-20 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547067
  • Size 100µg
Applications
Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytokine Zcyto10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547067 349.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MP700
  • Size 500ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; interferon gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MP700 349.00
Check your price
  • Phospho-HDAC5 (Ser498) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536647
  • Size 100µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536647 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530928
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530928 349.00
Check your price
  • CD64 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # CD6404
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6404 263.00
Check your price
  • CD64 Monoclonal Antibody, TRI-COLOR® conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate TRI-COLOR®
  • Catalog # CD6406
  • Size 0.5ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6406 321.00
Check your price
  • eIF4B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523286
  • Size 100ug
Applications
Western Blot (0.005-1 µg/ml)
Alternate names:
EIF-4B, PRO1843
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523286 349.00
Check your price
  • beta COP Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1061
  • Size 100µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (1:2,000), Western Blot (1:1,000)
Alternate names:
Beta-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1061 375.00
Check your price
  • COMP Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519850
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Thrombospondin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519850 349.00
Check your price
  • COMP Monoclonal Antibody for Western Blot, IHC (F), IHC (P), IP and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120221
  • Size 1mL
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Thrombospondin-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120221 349.00
Check your price
  • P-cadherin Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # 324000
  • Size 100µg
Applications
Immunofluorescence (2-10 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Placental cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
324000 305.00
Check your price
  • P-cadherin Monoclonal Antibody for Western Blot, IF, IHC (F), IP, ELISA and IA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 135800
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Placental cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
135800 330.00
Check your price