Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  117 
31 - 60 of 3500 products displayed
  • ARF6 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511795
  • Size 500µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Western Blot (1:100-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511795 349.00
Check your price
  • Rabbit IgG Isotype Control for IHC (P) and Ctrl
  • Host Rabbit
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # MA516384
  • Size 500µL
Applications
Control (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516384 499.00
Check your price
  • Rabbit IgG Isotype Control for IHC (P) and Ctrl
  • Host Rabbit
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # MA516385
  • Size 1mL
Applications
Control (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516385 700.00
Check your price
  • CDw17 Monoclonal Antibody for IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Many
  • Conjugate Unconjugated
  • Catalog # MA191271
  • Size 200µg
Applications
Flow Cytometry (1-5 µg for 10^6 cells), Immunocytochemistry (5-20 ug/ml), Immunofluorescence (5-20 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191271 349.00
Check your price
  • Mouse IgG1 Isotype Control, PE conjugate for Flow and Ctrl
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate PE
  • Catalog # MA518097
  • Size 100TESTS
Applications
Control (Assay-Dependent), Flow Cytometry (10ulx10^6 cells or 100ul whole blood)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518097 349.00
Check your price
  • Fetal Hemoglobin Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MHFH05
  • Size 500µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHFH05 484.00
Check your price
  • VGLL4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA545925
  • Size 100µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545925 309.00
Check your price
  • UBE4A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511955
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511955 349.00
Check your price
  • LAP (Latency Associated Peptide) Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517186
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517186 309.00
Check your price
  • TGF beta-1 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA515065
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515065 275.00
Check your price
  • PD-1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516134
  • Size 100µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516134 349.00
Check your price
  • MUC20 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514973
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514973 349.00
Check your price
  • Ki-67 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121520
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (10-20 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121520 399.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA191326
  • Size 500µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191326 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA513203
  • Size 500µL
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1-2 ug/ml), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513203 219.00
Check your price
  • AZGP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA544912
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544912 309.00
Check your price
  • APAF1 Monoclonal Antibody for IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515744
  • Size 100ug
Applications
Immunohistochemistry (0.5 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515744 349.00
Check your price
  • gamma Adaptin Monoclonal Antibody for Western Blot, IF, ICC, IP and IM
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MA125066
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunomicroscopy (1:100), Immunoprecipitation (1:100), Western Blot (1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125066 349.00
Check your price
  • RECA-1 Monoclonal Antibody for IF and IHC (F)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA181510
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181510 309.00
Check your price
  • Mouse IgG2b Isotype Control for Flow and Ctrl
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # PA533223
  • Size 100TESTS
Applications
Control (Assay-Dependent), Flow Cytometry (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533223 225.00
Check your price
  • Urease Polyclonal Antibody, HRP conjugate for Western Blot, ELISA and IM
  • Host Rabbit
  • Target Species Plant
  • Conjugate HRP
  • Catalog # PA128969
  • Size 1mg
Applications
ELISA (1:5000), Immunomicroscopy (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128969 309.00
Check your price
  • Mouse IgG2a Isotype Control, Alexa Fluor® 647 conjugate for Flow and Ctrl
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518170
  • Size 100µg
Applications
Control (Assay-dependent), Flow Cytometry (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518170 249.00
Check your price
  • ZNF683 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541242
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541242 309.00
Check your price
  • Vinculin Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700062
  • Size 100µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Western Blot (1-3µg/ml)
Alternate names:
Metavinculin
Catalog # Size Price (AUD) Your Price (AUD) Qty
700062 305.00
Check your price
  • TMEM65 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523864
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523864 349.00
Check your price
  • Synapsin 2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # OSS00020W
  • Size 100µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Synapsin II
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSS00020W 309.00
Check your price
  • SPRED1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520617
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520617 349.00
Check your price
  • Estriol Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175135
  • Size 100µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175135 600.00
Check your price
  • Estriol Polyclonal Antibody for RIA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA185737
  • Size 100µL
Applications
Radioimmune Assays (1:30,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185737 275.00
Check your price
  • mCherry Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534974
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:3000), Western Blot (1:1000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534974 349.00
Check your price