Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  117 
91 - 120 of 3500 products displayed
  • WT1 Monoclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA146028
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:400), Immunoprecipitation (2-10 µg/ml lysate), Western Blot (2 µg/ml)
Alternate names:
WT33
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146028 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 482300
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
482300 309.00
Check your price
  • Substance P Monoclonal Antibody for IF, ICC, IHC (F) and IHC (P)
  • Host Rat
  • Target Species Cat, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA517201
  • Size 500µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
PPT
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517201 309.00
Check your price
  • RAB5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA3915
  • Size 100µL
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1:500-1:5000)
Alternate names:
RAB5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3915 349.00
Check your price
  • Human Interleukin 13 (IL-13) for Western Blot, IP, ELISA and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RIL135
  • Size 5ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
interleukin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL135 293.00
Check your price
  • Cullin 2 Polyclonal Antibody for Western Blot, IHC (P), Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 511800
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Immunoprecipitation (2-3 µg/ml), Western Blot (1:250-1:1000)
Alternate names:
CUL-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
511800 307.00
Check your price
  • Clathrin Light Chain B Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127770
  • Size 50µg
Applications
ELISA (1 µg/ml), Western Blot (Assay Dependent)
Alternate names:
LCB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127770 425.00
Check your price
  • Citrulline Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA186258
  • Size 100µL
Applications
ELISA (1/1000-1/2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186258 1,000.00
Check your price
  • Normal Rat Serum for IF, ICC, IHC, BLOCK and Ctrl
  • Host Rat
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # 31888
  • Size 120mg
Applications
Blocking Assay (Assay Dependent), Control (Assay-Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31888 38.00
Check your price
  • Hepatocytes Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512417
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:20-1:40)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512417 309.00
Check your price
  • Tn Antigen Monoclonal Antibody for IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190544
  • Size 500µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (1:50-1:100), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190544 399.00
Check your price
  • Goat anti-Mouse IgG2b Secondary Antibody, RPE conjugate for IF and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate
  • Catalog # M32404
  • Size 500µL
Applications
Flow Cytometry (1:20-1:80)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32404 Not Orderable Online
Check your price
  • Goat anti-Mouse IgG2a Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # M32207
  • Size 1mL
Applications
ELISA (1:2,000), Immunohistochemistry (1:2,000), Western Blot (1:2,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32207 149.00
Check your price
  • Goat anti-Mouse IgG2a Secondary Antibody, Biotin conjugate for IF
  • Host Goat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # M32315
  • Size 500µL
Applications
Immunofluorescence (1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
M32315 115.00
Check your price
  • Rhodopsin Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1722
  • Size 100µL
Applications
Immunocytochemistry (1:1,000), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Opsin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1722 375.00
Check your price
  • PCNA Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Insect, Mouse, Non-human primate, Yeast
  • Conjugate Unconjugated
  • Catalog # 133900
  • Size 200µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Paraffin) (1:50-1:500), Immunoprecipitation (2-5 ug), Western Blot (0.1-1 ug/ml)
Alternate names:
Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
133900 308.00
Check your price
  • Oxytocin Receptor Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532961
  • Size 50ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (40 µg/ml)
Alternate names:
OT-R
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532961 349.00
Check your price
  • GFAP Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA135377
  • Size 1mL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin, Frozen) (1:10-1:50), Western Blot (Assay dependent)
Alternate names:
GFAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135377 309.00
Check your price
  • CD28 Monoclonal Antibody for Western Blot, ICC, IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110166
  • Size 100ug
Applications
Flow Cytometry (2-8 ug/ml), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
TP44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110166 159.00
Check your price
  • CD1d Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120223
  • Size 100µg
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-dependent)
Alternate names:
R3G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120223 375.00
Check your price
  • RAD51 Polyclonal Antibody for Western Blot, IP and ELISA
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA14924
  • Size 50µL
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
RAD51A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14924 425.00
Check your price
  • Normal Goat Serum for IF, ICC, IHC, BLOCK and Ctrl
  • Host Goat
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # 31873
  • Size 600mg
Applications
Blocking Assay (Assay Dependent), Control (Assay-Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31873 109.00
Check your price
  • Ebola Virus Monoclonal Antibody for Western Blot, ELISA and IM
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA121636
  • Size 500µg
Applications
ELISA (Assay dependent), Immunomicroscopy (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121636 475.00
Check your price
  • Nitrotyrosine Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # 321900
  • Size 100µg
Applications
Immunohistochemistry (1-2 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
321900 304.00
Check your price
  • Aspergillus Monoclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Fungi
  • Conjugate Unconjugated
  • Catalog # MA174434
  • Size 250µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1/300), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174434 475.00
Check your price
  • Neutrophils Monoclonal Antibody for IHC (F), IHC (P), Flow, ELISA and FN
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA140038
  • Size 100µg
Applications
ELISA (Assay Dependent.), Flow Cytometry (Assay Dependent.), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140038 449.00
Check your price
  • IL-3 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75595
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75595 2,600.00
Check your price
  • IL-2 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75589
  • Size 1mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75589 2,785.00
Check your price
  • Goat anti-Human IgG Fc Secondary Antibody for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A18819
  • Size 2mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18819 74.00
Check your price
  • TCR DO11.10 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517662
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517662 139.00
Check your price