Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  67 
1411 - 1440 of 2000 products displayed
  • GPI Antibody (Monoclonal, 1B7D7)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515396
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (Paraffin) (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Phosphohexose isomerase; Phosphoglucose isomerase; Neuroleukin; Autocrine motility factor; Sperm antigen 36
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515396 349.00
Check your price
  • GLDC Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526504
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Glycine dehydrogenase (aminomethyl-transferring); Glycine decarboxylase; Glycine cleavage system P protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526504 349.00
Check your price
  • DHX36 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534494
  • Size 100 ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
RNA helicase associated with AU-rich element ARE; MLE-like protein 1; DEAH box protein 36; G4-resolvase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534494 349.00
Check your price
  • CNDP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526174
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Serum carnosinase; Glutamate carboxypeptidase-like protein 2; Carnosine dipeptidase 1; CNDP dipeptidase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526174 349.00
Check your price
  • CD52 Antibody, FITC (Monoclonal, YTH34.5)
  • Host Rat
  • Target Species Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA517000
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
Human epididymis-specific protein 5; Epididymal secretory protein E5; Cambridge pathology 1 antigen; CDw52
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517000 449.00
Check your price
  • Calreticulin Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA3900
  • Size 100 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3900 375.00
Check your price
  • Clostridium perfringens Toxin B/C Antibody
  • Host Sheep
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA185311
  • Size 500 µL
Applications
ELISA (Assay Dependent)
Alternate names:
C. perfringens, C. perfringens Toxin B, C. perfringens Toxoid B, pCPF5603_62
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185311 349.00
Check your price
  • HSV Type 1 Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA17496
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17496 309.00
Check your price
  • TAGLN Antibody
  • Host Goat
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA518252
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Smooth muscle protein 22-alpha; Actin-associated protein p27; Protein WS3-10; 22 kDa actin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518252 349.00
Check your price
  • SARM Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA130598
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Tir-1 homolog; Sterile alpha motif domain-containing protein 2; Sterile alpha and Armadillo repeat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130598 399.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141089
  • Size 200 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141089 309.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527617
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527617 349.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534737
  • Size 100 µL
Applications
Gel Shift (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534737 349.00
Check your price
  • EPCR Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547382
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Endothelial cell protein C receptor; Centrosomal protein CCD41; Centrocyclin; Activated protein C receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547382 349.00
Check your price
  • PARP (Cleaved Asp214, Asp215) Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44698G
  • Size 100 µL
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
Poly[ADP-ribose] synthase 1; NAD(+) ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44698G 303.00
Check your price
  • CD204 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522956
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II; Scavenger receptor class A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522956 349.00
Check your price
  • MRPL3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521892
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MRL3, RPML3, 39S ribosomal protein L3, mitochondrial, L3mt, MRP-L3, mitochondrial 60S ribosomal protein L3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521892 349.00
Check your price
  • KLHL41 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540849
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Sarcosin; Kelch-related protein 1; Kelch repeat and BTB domain-containing protein 10; Kel-like protein 23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540849 309.00
Check your price
  • GLG1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532779
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
Golgi sialoglycoprotein MG-160; E-selectin ligand 1; Cysteine-rich fibroblast growth factor receptor; CFR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532779 349.00
Check your price
  • CDK5RAP3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538872
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein HSF-27; LXXLL/leucine-zipper-containing ARF-binding protein; CDK5 activator-binding protein C53
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538872 309.00
Check your price
  • HSV Type 1 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA129210
  • Size 6 mL
Applications
Immunohistochemistry (Paraffin) (Neat)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129210 309.00
Check your price
  • Nhe-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520444
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Solute carrier family 9 member 1; Na(+)/H(+) exchanger 1; Na(+)/H(+) antiporter, amiloride-sensitive; APNH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520444 349.00
Check your price
  • PABP Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527443
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
PAB1, PABP, PABP1, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 2, polyadenylate-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527443 349.00
Check your price
  • MIOX Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530650
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal-specific oxidoreductase; Myo-inositol oxygenase; Kidney-specific protein 32; Aldehyde reductase-like 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530650 349.00
Check your price
  • Phospho-SHIP1 (Tyr1021) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538393
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
p150Ship; SH2 domain-containing inositol 5'-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538393 309.00
Check your price
  • HADH Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528203
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Short-chain 3-hydroxyacyl-CoA dehydrogenase; Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528203 349.00
Check your price
  • Calreticulin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534786
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534786 349.00
Check your price
  • TRPM4 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00088W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Melastatin-like 2; MLS2s; Long transient receptor potential channel 4; Calcium-activated non-selective cation channel 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00088W 375.00
Check your price
  • SUZ12 Antibody (Monoclonal, 2C11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515732
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Suppressor of zeste 12 protein homolog; Joined to JAZF1 protein; Chromatin precipitated E2F target 9 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515732 349.00
Check your price
  • Phospho-SHC (Tyr239, Tyr240) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44830
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.5-2 µg/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
44830 309.00
Check your price