Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
391 - 420 of 2000 products displayed
  • Ankyrin B Antibody (Monoclonal, 2.2)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 333700
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (0.5-1 µg/ml)
Alternate names:
Brain ankyrin; Ankyrin-B; Non-erythroid ankyrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
333700 304.00
Check your price
  • AP2 gamma Antibody (Monoclonal, AP2g 6E4/4)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA191374
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Activating enhancer-binding protein 2 gamma; AP-2.2; Transcription factor ERF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191374 399.00
Check your price
  • APIP Antibody (Monoclonal, 19F424)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141380
  • Size 100 µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
APAF1-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141380 309.00
Check your price
  • Apolipoprotein A1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520721
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Apolipoprotein A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520721 349.00
Check your price
  • Apolipoprotein H Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537839
  • Size 100 µg
Applications
Western Blot (0.2-0.6 µg/ml)
Alternate names:
Beta-2-glycoprotein I; Apolipoprotein H; Anticardiolipin cofactor; Activated protein C-binding protein; APC inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537839 309.00
Check your price
  • ARF4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539010
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
ADP-ribosylation factor 4, ARF2, ADP-ribosylation factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539010 309.00
Check your price
  • Arylsulfatase A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547330
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cerebroside-sulfatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547330 349.00
Check your price
  • ASGR1 Antibody (Monoclonal, 8D7)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA140244
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
Hepatic lectin 1; Hepatic lectin H1; C-type lectin domain family 4 member H1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140244 600.00
Check your price
  • ATGL Antibody (Monoclonal, 494702)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524202
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Transport-secretion protein 2; TTS2.2; Pigment epithelium-derived factor; IPLA2-zeta; Desnutrin; Calcium-independent phospholipase A2; Adipose triglyceride lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524202 275.00
Check your price
  • ATP1A1 Antibody (Monoclonal, M8-P1-A3)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3929
  • Size 100 µg
Applications
Flow Cytometry (1/20-1/100), Immunocytochemistry (1/50-1/200), Immunofluorescence (1/50-1/200), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1/100-1/2000), Western Blot (1:200-1:2000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3929 375.00
Check your price
  • ATPIF1 Antibody (Monoclonal, 5E2D7)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # A21355
  • Size 100 µg
Applications
Western Blot (1.0 ug/ml)
Alternate names:
Inhibitor of F(1)F(o)-ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21355 327.00
Check your price
  • ATRX Antibody (Monoclonal, 5B3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518469
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Znf-HX; X-linked nuclear protein; X-linked helicase II; ATP-dependent helicase ATRX
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518469 309.00
Check your price
  • ATXN7L1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539033
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Ataxin-7-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539033 309.00
Check your price
  • Aurora A Antibody (Monoclonal, 35C1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA180399
  • Size 100 µg
Applications
ChIP assay (1-5µg), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1/500-1/1000)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; Serine/threonine-protein kinase 6; Ipl1- and aurora-related kinase 1; Aurora/IPL1-related kinase 1; Aurora family kinase 1; Aurora 2; Serine/threonine-protein kinase 15; Breast tumor-amplified kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180399 399.00
Check your price
  • BACE2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1753
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Western Blot (2 µg/ml)
Alternate names:
Theta-secretase; Membrane-associated aspartic protease 1; Memapsin-1; Beta-site amyloid precursor protein cleaving enzyme 2; Aspartyl protease 1; Down region aspartic protease; Aspartic-like protease 56 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1753 349.00
Check your price
  • Bacillus anthracis Protective Antigen Antibody (Monoclonal, BAP0101)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA121675
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
B. anthracis PA, Bacillus anthracis PA, B. anthracis Protective Antigen, Anthrax Protective Antigen, Anthrax PA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121675 349.00
Check your price
  • BAGE4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539049
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Cancer/testis antigen 2.4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539049 309.00
Check your price
  • beta Actin Antibody (Monoclonal, AC-15)
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Sheep, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # AM4302
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
AM4302 268.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116889
  • Size 100 µg
Applications
Western Blot (1:5000-1:15,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116889 375.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1183
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1183 225.00
Check your price
  • beta Amyloid (1-20) Antibody (Monoclonal, 7N22)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # AHB0272
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHB0272 361.00
Check your price
  • beta Amyloid (1-40) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44136
  • Size 100 µg
Applications
Dot blot (Assay Dependent), ELISA (1µg/ml), Immunohistochemistry (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI; ABPP
Catalog # Size Price (AUD) Your Price (AUD) Qty
44136 355.00
Check your price
  • beta Amyloid Antibody (Monoclonal, H31L21)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700254
  • Size 100 µg
Applications
ELISA (0.1-0.2 ug/ml), Immunohistochemistry (Paraffin) (1 ug/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
700254 307.00
Check your price
  • beta Arrestin 1 Antibody (Monoclonal, ZA005)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 395000
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (Assay Dependent)
Alternate names:
Arrestin beta-1; Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
395000 305.00
Check your price
  • beta Arrestin 1 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519582
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519582 349.00
Check your price
  • beta Catenin Antibody (Monoclonal, 6F9)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1300
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:50-1:1000), Immunohistochemistry (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:1000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1300 275.00
Check your price
  • beta Catenin Antibody (Monoclonal, CAT-5H10)
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 138400
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
138400 308.00
Check your price
  • beta Catenin Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 712700
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
712700 305.00
Check your price
  • beta-3 Tubulin Antibody (Monoclonal, 2 28 33)
  • Host Mouse
  • Target Species Dog, C. elegans, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 322600
  • Size 100 µg
Applications
Immunocytochemistry (1-3 µg/ml), Immunofluorescence (1-3 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Tubulin beta-III; Tubulin beta-4 chain; Neuron-specific class III beta-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
322600 307.00
Check your price
  • beta-3 Tubulin Antibody (Monoclonal, 2G10)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1118
  • Size 100 µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (5 µg), Western Blot (1:500-1:2000)
Alternate names:
Tubulin beta-III; Tubulin beta-4 chain; Neuron-specific class III beta-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1118 349.00
Check your price