Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
421 - 450 of 2000 products displayed
  • B7-H4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518771
  • Size 100 µg
Applications
Western Blot (0.5-1.5µg/ml)
Alternate names:
T-cell costimulatory molecule B7x; Protein B7S1; Immune costimulatory protein B7-H4; B7h.5; B7 homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518771 349.00
Check your price
  • BACE1 Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1177
  • Size 100 µg
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:1000-1:2000)
Alternate names:
Membrane-associated aspartic protease 2; Memapsin-2; Beta-site amyloid precursor protein cleaving enzyme 1; Aspartyl protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1177 309.00
Check your price
  • BACE1 Polyclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1757
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunoprecipitation (Assay dependent), Western Blot (1:200-1:2000)
Alternate names:
Membrane-associated aspartic protease 2; Memapsin-2; Beta-site amyloid precursor protein cleaving enzyme 1; Aspartyl protease 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1757 375.00
Check your price
  • BAFF Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701244
  • Size 100 µg
Applications
Western Blot (2.5-5 ug/ml)
Alternate names:
TNF- and APOL-related leukocyte expressed ligand 1; Dendritic cell-derived TNF-like molecule; BAFF; B-cell-activating factor; B lymphocyte stimulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
701244 309.00
Check your price
  • BCMA Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547794
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
B-cell maturation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547794 349.00
Check your price
  • beta Actin Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Sheep, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # AM4302
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
AM4302 268.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116889
  • Size 100 µg
Applications
Western Blot (1:5000-1:15,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116889 375.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1183
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1183 225.00
Check your price
  • beta Amyloid Monoclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700254
  • Size 100 µg
Applications
ELISA (0.1-0.2 ug/ml), Immunohistochemistry (Paraffin) (1 ug/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
700254 307.00
Check your price
  • beta Amyloid Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520737
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520737 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519582
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Arrestin beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519582 349.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA12001
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Immunoprecipitation (1 ug/ml), Western Blot (1-3 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12001 275.00
Check your price
  • beta Catenin Polyclonal Antibody for IF, ICC, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 712700
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
712700 305.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516308
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308 225.00
Check your price
  • beta-3 Tubulin Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Dog, C. elegans, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 322600
  • Size 100 µg
Applications
Immunocytochemistry (1-3 µg/ml), Immunofluorescence (1-3 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Tubulin beta-III; Tubulin beta-4 chain; Neuron-specific class III beta-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
322600 307.00
Check your price
  • Blood Group Lewis A Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119344
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Fucosyltransferase III; Fucosyltransferase 3; Blood group Lewis alpha-4-fucosyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119344 199.00
Check your price
  • BMP-6 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547695
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.75-3.75 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
VG-1-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547695 349.00
Check your price
  • Bovine RSV Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123354
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
BRSV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123354 349.00
Check your price
  • Btk Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520085
  • Size 100 µg
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Bruton tyrosine kinase; B-cell progenitor kinase; Agammaglobulinemia tyrosine kinase; Kinase EMB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520085 349.00
Check your price
  • c-Myc Tag Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow, IP, ChIP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1980
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay dependent), Flow Cytometry (1:50-1:200), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin, Frozen) (1:50-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:2000)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1980 249.00
Check your price
  • C-Peptide Monoclonal Antibody for ICC, IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119159
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (2-5 µg/ml), Radioimmune Assays (Assay Dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119159 309.00
Check your price
  • C/EBP beta Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123319
  • Size 100 µg
Applications
Immunohistochemistry (Assay-Dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123319 375.00
Check your price
  • Calnexin Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519169
  • Size 100 µg
Applications
ELISA (1-2 µg/ml), Immunohistochemistry (3-6 µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
p90; Major histocompatibility complex class I antigen-binding protein p88; IP90
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519169 349.00
Check your price
  • Canine Distemper Virus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122748
  • Size 100 µg
Applications
ELISA (Assay dependent)
Alternate names:
CDV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122748 309.00
Check your price
  • Canine Distemper Virus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122747
  • Size 100 µg
Applications
ELISA (Assay dependent)
Alternate names:
CDV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122747 309.00
Check your price
  • Canine Parvovirus Monoclonal Antibody for ELISA and IDF
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA122685
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay-dependent)
Alternate names:
CPV, parvo
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122685 309.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700182
  • Size 100 µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-0.2 ug/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
700182 309.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112505
  • Size 100 µg
Applications
Western Blot (0.25-0.5µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112505 325.00
Check your price
  • Caveolin 1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1064
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (5 µg), Western Blot (1-2 µg/ml)
Alternate names:
Vesicular integral-membrane protein VIP21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1064 375.00
Check your price
  • Caveolin 3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1066
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (1:100-200), Immunoprecipitation (5 µg), Western Blot (1-3 µg/ml)
Alternate names:
M-caveolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1066 375.00
Check your price