Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  40  50  60  70  80  90  100  110  117 
511 - 540 of 3500 products displayed
  • Calbindin D28K Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546936
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Vitamin D-dependent calcium-binding protein, avian-type; D-28K; Calbindin D28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546936 375.00
Check your price
  • Calcitonin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120285
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Calcitonin gene-related peptide I; Alpha-type CGRP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120285 309.00
Check your price
  • Calcium Channel 1H Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate Unconjugated
  • Catalog # PA539530
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav3.2; Low-voltage-activated calcium channel alpha1 3.2 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539530 309.00
Check your price
  • Calcium Sensing Receptor Monoclonal Antibody for Western Blot, IF, IHC (F) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1934
  • Size 100µg
Applications
ELISA (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (2 ug/ml), Western Blot (2 ug/ml)
Alternate names:
Parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1934 375.00
Check your price
  • Calreticulin Monoclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Hamster, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA191034
  • Size 100µg
Applications
Flow Cytometry (1:100), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (2 µg/ml), Immunoprecipitation (12.5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
grp60; HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191034 475.00
Check your price
  • CaMKII beta/gamma/delta Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539732
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539732 309.00
Check your price
  • CaMKII delta Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518532
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
CAMK2B, CAMK2D, CAMK2G, Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMKG, Calcium/calmodulin-dependent protein kinase type II subunitdelta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518532 309.00
Check your price
  • Candida albicans Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Fungi, Yeast
  • Conjugate Unconjugated
  • Catalog # MA518176
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
C. albicans
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518176 309.00
Check your price
  • Canine Distemper Virus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17468
  • Size 100µg
Applications
ELISA (1:20)
Alternate names:
CDV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17468 309.00
Check your price
  • Carbonic Anhydrase IV Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547312
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.25-12.5 µg/ml)
Alternate names:
Carbonic anhydrase IV; Carbonate dehydratase IV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547312 349.00
Check your price
  • Caspase 9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112506
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112506 375.00
Check your price
  • Cathepsin D Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547046
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
OTTHUMP00000196039, OTTHUMP00000198692, ceroid-lipofuscinosis, neuronal 10, lysosomal aspartyl peptidase, lysosomal aspartyl protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547046 349.00
Check your price
  • Cathepsin S Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547088
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cathepsin S, CTSS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547088 349.00
Check your price
  • Cathepsin V Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523928
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cathepsin V; Cathepsin U
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523928 275.00
Check your price
  • Caveolin 1 Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1064
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (5 µg), Western Blot (1-2 µg/ml)
Alternate names:
Vesicular integral-membrane protein VIP21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1064 375.00
Check your price
  • Caveolin 3 Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1066
  • Size 100µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (1:100-200), Immunoprecipitation (5 µg), Western Blot (1-3 µg/ml)
Alternate names:
M-caveolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1066 375.00
Check your price
  • CCL3 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701097
  • Size 100µg
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701097 309.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, IHC (F), ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546951
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.05-0.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546951 375.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121621
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunocytochemistry (1:800), Immunofluorescence (1:800), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121621 425.00
Check your price
  • CD103 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121434
  • Size 100µg
Applications
Flow Cytometry (2 µg for 5x10^5 cells), Immunohistochemistry (Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121434 349.00
Check your price
  • CD103 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547236
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547236 349.00
Check your price
  • CD109 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547620
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
r150; p180; Platelet-specific Gov antigen; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; 150 kDa TGF-beta-1-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547620 349.00
Check your price
  • CD11b Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120145
  • Size 100µg
Applications
ELISA (1/1000)
Alternate names:
Neutrophil adherence receptor; Leukocyte adhesion receptor MO1; Cell surface glycoprotein MAC-1 subunit alpha; CR-3 alpha chain; CD11 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120145 349.00
Check your price
  • CD11c Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA182105
  • Size 100µg
Applications
Flow Cytometry (Neat- 1/10)
Alternate names:
Leukocyte adhesion receptor p150,95; Leukocyte adhesion glycoprotein p150,95 alpha chain; Leu M5; CD11 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182105 349.00
Check your price
  • CD127 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547443
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
CDw127
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547443 349.00
Check your price
  • CD14 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA119514
  • Size 100µg
Applications
Flow Cytometry (1:1000)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119514 309.00
Check your price
  • CD14 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Dog, Camelid, Cat, Goat, Human, Mink, Non-human primate, Sheep, Pig, Rabbit
  • Conjugate FITC
  • Catalog # MA182074
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182074 325.00
Check your price
  • CD142 Monoclonal Antibody for Western Blot, IF, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA183495
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Thromboplastin; Coagulation factor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183495 309.00
Check your price
  • CD146 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 357800
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (0.5-2 ug/ml)
Alternate names:
S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Melanoma cell adhesion molecule; Cell surface glycoprotein P1H12; Gicerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
357800 309.00
Check your price
  • CD147 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA135092
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Membrane glycoprotein gp42; HT7 antigen; Basic immunoglobulin superfamily; Neurothelin; Blood-brain barrier HT7 antigen; 5A11 antigen; OX-47 antigen; Glycoprotein CE9; Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135092 309.00
Check your price