Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  40  50  60  70  80  90  100  110  117 
541 - 570 of 3500 products displayed
  • beta-Arrestin 2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA1732
  • Size 100µg
Applications
Western Blot (1:2000)
Alternate names:
Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1732 349.00
Check your price
  • betacellulin Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547044
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (0.1-0.4 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
BTC, Probetacellulin, Betacellulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547044 375.00
Check your price
  • betacellulin Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546928
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.04-0.08 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
BTC, Probetacellulin, Betacellulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546928 375.00
Check your price
  • Blood Group A Antigen Monoclonal Antibody for IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191429
  • Size 100µg
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191429 375.00
Check your price
  • BMP-1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547222
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Procollagen C-proteinase; Mammalian tolloid protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547222 349.00
Check your price
  • BMP-6 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547695
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.75-3.75 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
VG-1-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547695 349.00
Check your price
  • BMPR1A Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 386000
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase receptor R5; BMP-2/BMP-4 receptor; Activin receptor-like kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
386000 305.00
Check your price
  • BNIP3L Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 393300
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NIP3-like protein X; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; Adenovirus E1B19K-binding protein B5
Catalog # Size Price (AUD) Your Price (AUD) Qty
393300 304.00
Check your price
  • Bovine RSV Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123355
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
BRSV
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123355 375.00
Check your price
  • BRCA1 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123162
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1-10 µg/ml)
Alternate names:
RING finger protein 53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123162 375.00
Check your price
  • BrdU Monoclonal Antibody for ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # OMA106200
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
OMA106200 375.00
Check your price
  • BrdU Monoclonal Antibody, FITC conjugate for IHC and Flow
  • Host Mouse
  • Target Species Chemical
  • Conjugate FITC
  • Catalog # MA112686
  • Size 100µg
Applications
Flow Cytometry (1-5 µg/10^6 cells), Immunohistochemistry (Assay Dependent)
Alternate names:
5-Bromo-2-deoxyuridine, Bromodeoxyuridine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112686 309.00
Check your price
  • BST-2 Polyclonal Antibody for Western Blot, ELISA and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131166
  • Size 100µg
Applications
Dot blot (1:2000), ELISA (1:2000), Western Blot (1:500-1:1000)
Alternate names:
Tetherin; HM1.24 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131166 600.00
Check your price
  • BTN2A2 Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547670
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
BT2.2, BTF2, butyrophilin 2, butyrophilin subfamily 2 member A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547670 375.00
Check your price
  • c-Kit Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 348800
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; p145 c-kit; Tyrosine-protein kinase Kit; Proto-oncogene c-Kit; Piebald trait protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
348800 309.00
Check your price
  • c-Mpl Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547121
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Proto-oncogene c-Mpl; Myeloproliferative leukemia protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547121 349.00
Check your price
  • C-Peptide Monoclonal Antibody for ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122707
  • Size 100µg
Applications
ELISA (Assay dependent), Radioimmune Assays (Assay dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122707 349.00
Check your price
  • C-Reactive Protein (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75528
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75528 209.00
Check your price
  • CA125 Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190039
  • Size 100µg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (1-5 µg/ml), Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Ovarian carcinoma antigen CA125; Ovarian cancer-related tumor marker CA125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190039 375.00
Check your price
  • Calbindin D28K Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546936
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Vitamin D-dependent calcium-binding protein, avian-type; D-28K; Calbindin D28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546936 375.00
Check your price
  • Calcineurin B Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA523933
  • Size 100µg
Applications
Western Blot (0.5 µg/ml)
Alternate names:
Protein phosphatase 3 regulatory subunit B alpha isoform 1; Protein phosphatase 2B regulatory subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523933 275.00
Check your price
  • Calcium Channel gamma-3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112674
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Transmembrane AMPAR regulatory protein gamma-3; Neuronal voltage-gated calcium channel gamma-3 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112674 309.00
Check your price
  • CaMKII alpha Monoclonal Antibody for Western Blot, IHC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Fish, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # 137300
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, calcium/calmodulin-dependent protein kinase type II alpha chain, Calci
Catalog # Size Price (AUD) Your Price (AUD) Qty
137300 304.00
Check your price
  • Candida albicans Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Fungi, Yeast
  • Conjugate Unconjugated
  • Catalog # MA518176
  • Size 100µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
C. albicans
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518176 309.00
Check your price
  • Canine Heartworm Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518178
  • Size 100µg
Applications
ELISA (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518178 289.00
Check your price
  • Carbonic Anhydrase XII Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547303
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Carbonic anhydrase XII; Carbonate dehydratase XII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547303 349.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700182
  • Size 100µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-0.2 ug/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
700182 309.00
Check your price
  • Catalase Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518531
  • Size 100µg
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
CAT; catalase ; HGNC:1516; MGC138422; MGC138424
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518531 349.00
Check your price
  • Cathepsin Z Polyclonal Antibody for Western Blot, IF, ICC, IP and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547048
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cathepsin P, Cathepsin Z, CTSZ
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547048 375.00
Check your price
  • Caveolin 1 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 036000
  • Size 100µg
Applications
Immunofluorescence (2-4 ug/ml), Immunoprecipitation (2-5 ug/ml), Western Blot (0.5-2.0 ug/ml)
Alternate names:
Phospho-Caveolin-1 pTyr14
Catalog # Size Price (AUD) Your Price (AUD) Qty
036000 365.00
Check your price