Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  40  50  60  70  80  90  100  110  117 
571 - 600 of 3500 products displayed
  • Canine Heartworm Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Nematode
  • Conjugate Unconjugated
  • Catalog # MA518178
  • Size 100µg
Applications
ELISA (1:20-1:200)
Alternate names:
Adult canine heartworm, Filaria, dirofilariasis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518178 289.00
Check your price
  • Carbonic Anhydrase XII Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547303
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Carbonic anhydrase XII; Carbonate dehydratase XII
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547303 349.00
Check your price
  • Caspase 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700182
  • Size 100µg
Applications
Flow Cytometry (0.5-1 ug/test), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-0.2 ug/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain; LICE
Catalog # Size Price (AUD) Your Price (AUD) Qty
700182 309.00
Check your price
  • Catalase Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518531
  • Size 100µg
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
CAT; catalase ; HGNC:1516; MGC138422; MGC138424
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518531 349.00
Check your price
  • Cathepsin L Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547971
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Major excreted protein; Cathepsin L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547971 349.00
Check your price
  • CCR5 Monoclonal Antibody for IHC (P), Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124666
  • Size 100µg
Applications
Flow Cytometry (5-10 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Neutralization (Assay-dependent)
Alternate names:
HIV-1 fusion coreceptor; CHEMR13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124666 349.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519777
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519777 349.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121621
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunocytochemistry (1:800), Immunofluorescence (1:800), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121621 425.00
Check your price
  • CCRD6 Polyclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121614
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121614 399.00
Check your price
  • CD103 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121434
  • Size 100µg
Applications
Flow Cytometry (2 µg for 5x10^5 cells), Immunohistochemistry (Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121434 349.00
Check your price
  • CD103 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547236
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547236 349.00
Check your price
  • CD105 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523894
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Frozen) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523894 275.00
Check your price
  • CD109 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547620
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
r150; p180; Platelet-specific Gov antigen; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; 150 kDa TGF-beta-1-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547620 349.00
Check your price
  • CD11a Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519161
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (4-6µg/ml), Western Blot (0.5-2µg/ml)
Alternate names:
Leukocyte function-associated molecule 1 alpha chain; Leukocyte adhesion glycoprotein LFA-1 alpha chain; CD11 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519161 349.00
Check your price
  • CD13 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112033
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp150; Myeloid plasma membrane glycoprotein CD13; Microsomal aminopeptidase; Aminopeptidase M; Alanyl aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112033 225.00
Check your price
  • CD142 Monoclonal Antibody for Western Blot, IF, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA183495
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Thromboplastin; Coagulation factor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183495 309.00
Check your price
  • CD142 Polyclonal Antibody for Western Blot, Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547465
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Coagulation factor III
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547465 349.00
Check your price
  • CD163 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA180112
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD163 antigen, hemoglobin scavenger receptor, macrophage-associated antigen, scavenger receptor cysteine-rich type 1 protein M130
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180112 499.00
Check your price
  • CD166 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523850
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Activated leukocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523850 275.00
Check your price
  • CD1d Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120223
  • Size 100µg
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-dependent)
Alternate names:
R3G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120223 375.00
Check your price
  • CD22 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A15388
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15388 244.00
Check your price
  • CD229 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524034
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Signaling lymphocytic activation molecule 3; SLAM family member 3; Lymphocyte antigen 9; Cell surface molecule Ly-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524034 275.00
Check your price
  • CD27 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119245
  • Size 100µg
Applications
Flow Cytometry (2µg/ml)
Alternate names:
Tumor necrosis factor receptor superfamily member 7; T14; T-cell activation antigen CD27; CD27L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119245 225.00
Check your price
  • CD300e Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523940
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Polymeric immunoglobulin receptor 2; Immune receptor expressed on myeloid cells 2; CMRF35-A5; CD300 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523940 275.00
Check your price
  • CD320 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547155
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Transcobalamin receptor; FDC-signaling molecule 8D6; 8D6 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547155 349.00
Check your price
  • CD34 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547849
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547849 349.00
Check your price
  • CD34 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547582
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547582 349.00
Check your price
  • CD38 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119523
  • Size 100µg
Applications
Flow Cytometry (1:500)
Alternate names:
T10; Cyclic ADP-ribose hydrolase 1; ADP-ribosyl cyclase 1; 2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119523 225.00
Check your price
  • CD41 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA182655
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Platelet membrane glycoprotein IIb; GPalpha IIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182655 199.00
Check your price
  • CD45 Monoclonal Antibody, APC-Cyanine7 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC-Cyanine7
  • Catalog # A15395
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15395 204.00
Check your price