Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  50  60  67 
751 - 780 of 2000 products displayed
  • Spinesin Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546992
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Spinesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546992 375.00
Check your price
  • SPOCK1 Antibody (Monoclonal, 316801)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524039
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Protein SPOCK
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524039 275.00
Check your price
  • SRC3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1845
  • Size 100 µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Thyroid hormone receptor activator molecule 1; Steroid receptor coactivator protein 3; Receptor-associated coactivator 3; Class E basic helix-loop-helix protein 42; CBP-interacting protein; Amplified in breast cancer 1 protein; ACTR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1845 375.00
Check your price
  • Stabilin 2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547522
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Hyaluronan receptor for endocytosis; Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavenger receptor 2; FAS1 EGF-like and X-link domain-containing adhesion molecule 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547522 349.00
Check your price
  • SULF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112860
  • Size 100 µg
Applications
Western Blot (0.05-0.1 µg/ml)
Alternate names:
RP5-1049G16.1, DKFZp313E091, FLJ44488, FLJ90554, HSULF-2, KIAA1247, MGC126411
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112860 399.00
Check your price
  • Tau Antibody (Monoclonal, TAU-5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # AHB0042
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHB0042 311.00
Check your price
  • TGN46 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA11069
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (1:250 to 1:500)
Alternate names:
Trans-Golgi network protein TGN51; TGN48; TGN46; TGN38 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11069 349.00
Check your price
  • Thyroxine Antibody (Monoclonal, XM212)
  • Host Mouse
  • Target Species Chemical, Mammal
  • Conjugate Unconjugated
  • Catalog # MA122093
  • Size 100 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122093 309.00
Check your price
  • TLR3 Antibody
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA520183
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520183 349.00
Check your price
  • TLR4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 482300
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
482300 309.00
Check your price
  • TMEFF1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547998
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Transmembrane protein with EGF-like and one follistatin-like domain; M7365
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547998 309.00
Check your price
  • TMEM27 Antibody
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547676
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Transmembrane protein 27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547676 349.00
Check your price
  • TNF alpha Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519810
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519810 349.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, H68.4)
  • Host Mouse
  • Target Species Chicken, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 136800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
136800 312.00
Check your price
  • TRIM46 Antibody (Monoclonal, 3G11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522628
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml)
Alternate names:
Tripartite, fibronectin type-III and C-terminal SPRY motif protein; Gene Y protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522628 309.00
Check your price
  • TRIP15 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14139
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Thyroid receptor-interacting protein 15; JAB1-containing signalosome subunit 2; Alien homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14139 309.00
Check your price
  • TROP2 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547074
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cell surface glycoprotein Trop-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547074 375.00
Check your price
  • Trypsin 2 Antibody (Monoclonal, 410322)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524148
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Trypsin II; Serine protease 2; Anionic trypsinogen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524148 275.00
Check your price
  • TSPAN8 Antibody (Monoclonal, 657909)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524296
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (8-25 µg/ml)
Alternate names:
TSPAN8, tetraspanin 8, CO-029, TM4SF3, transmembrane 4 superfamily member 3, tumor-associated antigen CO-029
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524296 275.00
Check your price
  • TXNL5 Antibody (Monoclonal, 544710)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524245
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Thioredoxin-like protein 5; Protein 42-9-9; 14 kDa thioredoxin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524245 275.00
Check your price
  • Ubiquitin Antibody (Monoclonal, Ubi-1)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # 131600
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (2-5 ug/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Ubiquitin-like product Chap1/Dsk2; Protein linking IAP with cytoskeleton 2; DSK2 homolog; Chap1
Catalog # Size Price (AUD) Your Price (AUD) Qty
131600 306.00
Check your price
  • Varicella zoster Virus Antibody (Monoclonal, SG3)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA190605
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Varicellovirus, VZV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190605 309.00
Check your price
  • VDR Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186623
  • Size 100 µg
Applications
ELISA (1:16,000), Western Blot (0.3-1 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186623 375.00
Check your price
  • WNT7A/WNT7B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547430
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
protein Wnt-7a, proto-oncogene Wnt7a protein, protein Wnt-7b, proto-oncogene Wnt7b protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547430 349.00
Check your price
  • XBP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520218
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520218 349.00
Check your price
  • Yersinia pestis F1 Antigen Antibody (Monoclonal, F81D)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17427
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
Bubonic Plague, Pasturella pestis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17427 309.00
Check your price
  • ZNF181 Antibody (Monoclonal, 5F1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523257
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
HHZ181
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523257 309.00
Check your price
  • ZNF274 Antibody (Monoclonal, 1D8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521159
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein zfp2; Zinc finger protein with KRAB and SCAN domains 19; Zinc finger protein HFB101; Zinc finger protein 274
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521159 309.00
Check your price
  • ZNF821 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538041
  • Size 100 µg
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (20 µg/ml), Western Blot (2.5 µg/ml)
Alternate names:
Zinc finger protein 821
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538041 309.00
Check your price
  • ZO-1 Antibody (Monoclonal, ZO1-1A12)
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated See Additional Formats Alexa Fluor 488 Alexa Fluor 594 FITC
  • Catalog # 339100
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
339100 311.00
Check your price