Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
211 - 240 of 2000 products displayed
  • ALDH9A1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519329
  • Size 100 ug
Applications
Immunohistochemistry (3-5 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
R-aminobutyraldehyde dehydrogenase; Gamma-aminobutyraldehyde dehydrogenase; Aldehyde dehydrogenase family 9 member A1; Aldehyde dehydrogenase E3 isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519329 349.00
Check your price
  • ALOX15 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519052
  • Size 100 ug
Applications
Western Blot (0.2-0.5 ug/ml)
Alternate names:
Arachidonate omega-6 lipoxygenase; Arachidonate 12-lipoxygenase, leukocyte-type; 12/15-lipoxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519052 349.00
Check your price
  • Anillin Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518199
  • Size 100 ug
Applications
Immunohistochemistry (3-10µg/ml)
Alternate names:
ANLN; Scraps; ANILLIN; anillin, actin binding protein (scraps homolog, Drosophila); anillin (Drosophila Scraps homolog), actin binding protein; anillin, actin binding protein; DKFZp779A055; scra
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518199 349.00
Check your price
  • ASK1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519880
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
MAPK/ERK kinase kinase 5; Apoptosis signal-regulating kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519880 349.00
Check your price
  • ATG101 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521000
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
PP894, ATG101; Atg13-interacting protein; autophagy-related protein 101
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521000 349.00
Check your price
  • Beclin 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520171
  • Size 100 ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Coiled-coil myosin-like BCL2-interacting protein; Protein GT197
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520171 349.00
Check your price
  • beta Galactosidase Antibody (Monoclonal, BG-02)
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA110057
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110057 225.00
Check your price
  • Blimp-1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518110
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Positive regulatory domain I-binding factor 1; PR domain-containing protein 1; Beta-interferon gene positive regulatory domain I-binding factor; BLIMP-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518110 349.00
Check your price
  • Blood Group A1, A2, A3 Antigen Antibody (Monoclonal, Z2B-1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17665
  • Size 100 ug
Applications
Agglutination (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17665 309.00
Check your price
  • BMPR1A Antibody (Monoclonal, 4B7B2)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517036
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Serine/threonine-protein kinase receptor R5; BMP-2/BMP-4 receptor; Activin receptor-like kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517036 309.00
Check your price
  • Bovine Serum Albumin Antibody
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA523403
  • Size 100 ug
Applications
Western Blot (0.02-0.1 µg/ml)
Alternate names:
BSA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523403 349.00
Check your price
  • C/EBP delta Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534466
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
C/EBP-related protein 3; Transcription factor CELF; Nuclear factor NF-IL6-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534466 349.00
Check your price
  • C1D Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519053
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
C1D; MGC12261; MGC14659; SUNCOR; C1D DNA-binding protein; small unique nuclear receptor corepressor; nuclear DNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519053 349.00
Check your price
  • Caspase 8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520119
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520119 349.00
Check your price
  • Caspase 8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520118
  • Size 100 ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
MORT1-associated ced-3 homolog; ICE-like apoptotic protease 5; FADD-like ICE; FADD-homologous ICE/ced-3-like protease; CAP4; Apoptotic protease Mch-5; Apoptotic cysteine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520118 349.00
Check your price
  • Cathepsin K Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518950
  • Size 100 ug
Applications
ELISA (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Cathepsin X; Cathepsin O2; Cathepsin O
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518950 349.00
Check your price
  • CD16 Antibody (Monoclonal, ASH 1975)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA17633
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; Fc-gamma RIII; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17633 309.00
Check your price
  • CD20 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19024
  • Size 100 ug
Applications
Western Blot (0.03-0.1 µg/ml)
Alternate names:
Membrane-spanning 4-domains subfamily A member 1; Leukocyte surface antigen Leu-16; Bp35; B-lymphocyte surface antigen B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19024 309.00
Check your price
  • CD206 Antibody, FITC (Monoclonal, MR5D3)
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA516870
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
AW259686, CD206, MR, MMR, macrophage mannose receptor 1, MRC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516870 309.00
Check your price
  • CD22 Antibody (Monoclonal, MYG13)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA17635
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell surface antigen Leu-14; Sialic acid-binding Ig-like lectin 2; B-lymphocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17635 309.00
Check your price
  • CD267 (TACI) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519972
  • Size 100 ug
Applications
Immunocytochemistry (2 µg/ml), Western Blot (5 µg/ml)
Alternate names:
Transmembrane activator and CAML interactor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519972 349.00
Check your price
  • CD4 Antibody (Monoclonal, 8G1B12)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515775
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
W3/25 antigen; T-cell surface antigen T4/Leu-3; T-cell differentiation antigen L3T4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515775 349.00
Check your price
  • CD4 Antibody (Monoclonal, B486A1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514760
  • Size 100 ug
Applications
Flow Cytometry (1/200-1/400)
Alternate names:
T-cell surface antigen T4/Leu-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514760 349.00
Check your price
  • CD45 Antibody (Monoclonal, EM-05)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA110231
  • Size 100 ug
Applications
Flow Cytometry (2 ug/ml), Immunocytochemistry (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110231 119.00
Check your price
  • CD45 Antibody, APC (Monoclonal, EM-05)
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA110232
  • Size 100 ug
Applications
Flow Cytometry (4 ug/ml)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110232 275.00
Check your price
  • CD45 Antibody, Biotin (Monoclonal, YKIX716.13)
  • Host Rat
  • Target Species Dog
  • Conjugate Biotin
  • Catalog # MA516996
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516996 499.00
Check your price
  • CK1 epsilon Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518704
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
CSNK1E; casein kinase 1, epsilon; RP1-5O6.1; HCKIE; MGC10398 ; CKI-epsilon; OTTHUMP00000063258; OTTHUMP00000063263; casein kinase 1 epsilon; casein kinase I epsilon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518704 349.00
Check your price
  • COMT Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518554
  • Size 100 ug
Applications
ELISA (0.5-2.0ug/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
COMT; catechol-O-methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518554 349.00
Check your price
  • Cortactin Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA19030
  • Size 100 ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); Cttn; CORTACTIN; oncogene EMS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19030 309.00
Check your price
  • CPT1B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518062
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Carnitine palmitoyltransferase I-like protein; Carnitine palmitoyltransferase 1B; Carnitine O-palmitoyltransferase I, muscle isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518062 349.00
Check your price