Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
241 - 270 of 2000 products displayed
  • COMT Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518554
  • Size 100 ug
Applications
ELISA (0.5-2.0ug/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
COMT; catechol-O-methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518554 349.00
Check your price
  • Cortactin Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA19030
  • Size 100 ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
EMS1; ems1 sequence (mammary tumor and squamous cell carcinoma-associated (p80/85 src substrate); Cttn; CORTACTIN; oncogene EMS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19030 309.00
Check your price
  • CPT1B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518062
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Carnitine palmitoyltransferase I-like protein; Carnitine palmitoyltransferase 1B; Carnitine O-palmitoyltransferase I, muscle isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518062 349.00
Check your price
  • CRABP2 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523213
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Cellular retinoic acid-binding protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523213 349.00
Check your price
  • CRFR1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518801
  • Size 100 ug
Applications
Immunocytochemistry (0.1-0.5ug/ml), Immunofluorescence (2ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Corticotropin-releasing hormone receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518801 349.00
Check your price
  • CRP Antibody, Biotin (Monoclonal, C7)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110323
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110323 349.00
Check your price
  • CTRP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520145
  • Size 100 ug
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Putative secreted protein ZSIG37; G protein-coupled receptor-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520145 349.00
Check your price
  • CYP3A4 Antibody (Monoclonal, 3H8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517064
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Taurochenodeoxycholate 6-alpha-hydroxylase; Quinine 3-monooxygenase; Nifedipine oxidase; Cytochrome P450-PCN1; Cytochrome P450 NF-25; Cytochrome P450 HLp; Cytochrome P450 3A3; Cholesterol 25-hydroxylase; CYPIIIA4; CYPIIIA3; Albendazole sulfoxidase; Albendazole monooxygenase; 1,8-cineole 2-exo-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517064 309.00
Check your price
  • Cytochrome C Antibody
  • Host Chicken
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19585
  • Size 100 ug
Applications
ELISA (1:2,000-1:10,000), Western Blot (1:2,000-1:10,000)
Alternate names:
cyt c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19585 309.00
Check your price
  • DCXR Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518071
  • Size 100 ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Sperm surface protein P34H; Short chain dehydrogenase/reductase family 20C member 1; Kidney dicarbonyl reductase; Dicarbonyl/L-xylulose reductase; Carbonyl reductase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518071 349.00
Check your price
  • DFFB Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519913
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
DNA fragmentation factor 40 kDa subunit; Caspase-activated nuclease; Caspase-activated deoxyribonuclease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519913 349.00
Check your price
  • DHX36 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534494
  • Size 100 ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/mL)
Alternate names:
RNA helicase associated with AU-rich element ARE; MLE-like protein 1; DEAH box protein 36; G4-resolvase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534494 349.00
Check your price
  • Dnmt2 Antibody (Monoclonal, 102B1259.2)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516197
  • Size 100 ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
Met-2; DNA methyltransferase homolog MmuIIP; DNA (cytosine-5)-methyltransferase-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516197 349.00
Check your price
  • DR5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519895
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (10 ug/ml), Western Blot (2 µg/ml)
Alternate names:
MK; Death receptor 5; TNF-related apoptosis-inducing ligand receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519895 349.00
Check your price
  • Drosha Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518613
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
p241; Ribonuclease III; Protein Drosha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518613 349.00
Check your price
  • DUSP16 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523140
  • Size 100 ug
Applications
ChIP assay (1-3 ul), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523140 349.00
Check your price
  • EBF1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518720
  • Size 100 ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.3-0.1 ug/ml)
Alternate names:
Early B-cell factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518720 349.00
Check your price
  • Enterovirus pan Antibody (Monoclonal, 6A9)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516316
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Coxsakievirus, Echovirus, Cox B2, Cox B3, Echo 20, Echo 9, Echovirus 9, Echovirus 20
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516316 349.00
Check your price
  • Ephrin A1 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519397
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Immediate early response protein B61; EPH-related receptor tyrosine kinase ligand 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519397 349.00
Check your price
  • ERRF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534534
  • Size 100 ug
Applications
Immunocytochemistry (25 ug/ml), Immunofluorescence (25 ug/ml), Immunohistochemistry (Assay Dependent), Western Blot (1 ug/mL)
Alternate names:
ERRF, Estrogen receptor-related factor, ER-related factor, RP11-5P18.4, C1orf64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534534 349.00
Check your price
  • ETEA Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518183
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
UBX domain-containing protein 8; UBX domain-containing protein 3B; Protein ETEA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518183 349.00
Check your price
  • FANCG Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518203
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
DNA repair protein XRCC9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518203 349.00
Check your price
  • FGF4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520483
  • Size 100 ug
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Transforming protein KS3; Heparin-binding growth factor 4; Heparin secretory-transforming protein 1; K-fibroblast growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520483 349.00
Check your price
  • FKBP15 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520730
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
WASP and FKBP-like; 133 kDa FK506-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520730 349.00
Check your price
  • Follicle Stimulating Hormone Antibody (Monoclonal, 1G6D12)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514726
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Follitropin beta chain; Follicle-stimulating hormone beta subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514726 349.00
Check your price
  • FOXQ1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518178
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Hepatocyte nuclear factor 3 forkhead homolog 1; HNF-3/forkhead-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518178 349.00
Check your price
  • FZD7 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518883
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
FzE3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518883 349.00
Check your price
  • GABARAP Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534559
  • Size 100 ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
GABA(A) receptor-associated protein; MM46
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534559 349.00
Check your price
  • GAPDH Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # TAB1001
  • Size 100 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH
Catalog # Size Price (AUD) Your Price (AUD) Qty
TAB1001 249.00
Check your price
  • Gemin 2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520978
  • Size 100 ug
Applications
Immunocytochemistry (4 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Survival of motor neuron protein-interacting protein 1; Component of gems 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520978 349.00
Check your price