1 - 14 of 14 products displayed
  • Phospho-VDR (Ser51) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539757
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539757 309.00
Check your price
  • VDR Monoclonal Antibody for Western Blot, IF, ICC, IP, ChIP and GS
  • Host Rat
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA1710
  • Size 100 µg
Applications
ChIP assay (Assay dependent), Gel Shift (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1710 375.00
Check your price
  • VDR Monoclonal Antibody, Biotin conjugate for Western Blot and GS
  • Host Rat
  • Target Species Amphibian, Chicken, Fish, Hamster, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA514614
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514614 399.00
Check your price
  • VDR Monoclonal Antibody for Western Blot, IF, ICC, Flow and GS
  • Host Rat
  • Target Species Pig, Rat, Amphibian, Chicken, Fish, Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514617
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Western Blot (1-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514617 349.00
Check your price
  • Phospho-VDR (Ser208) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538931
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538931 309.00
Check your price
  • VDR Polyclonal Antibody for Western Blot, IP and GS
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1711
  • Size 100 µL
Applications
Gel Shift (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:100)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1711 375.00
Check your price
  • VDR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19116
  • Size 100 ug
Applications
Western Blot (0.3-1.0 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19116 309.00
Check your price
  • VDR Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186623
  • Size 100 µg
Applications
ELISA (1:16,000), Western Blot (0.3-1 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186623 375.00
Check your price
  • VDR Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535156
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535156 349.00
Check your price
  • CYP24A1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519406
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Cytochrome P450-CC24; Cytochrome P450 24A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519406 349.00
Check your price
  • CYP24A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521704
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytochrome P450-CC24; Cytochrome P450 24A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521704 349.00
Check your price
  • TXNIP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523485
  • Size 100 ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
Vitamin D3 up-regulated protein 1; Thioredoxin-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523485 349.00
Check your price
  • TXNIP Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # 403700
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vitamin D3 up-regulated protein 1; Thioredoxin-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
403700 304.00
Check your price
  • TXNIP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # 404600
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Vitamin D3 up-regulated protein 1; Thioredoxin-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
404600 304.00
Check your price