1 - 6 of 6 products displayed
  • E-Selectin/P-Selectin Monoclonal Antibody for Western Blot, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA180430
  • Size 200µg
Applications
ELISA (10 ug/ml), Flow Cytometry (10 ug/ml), Immunohistochemistry (Frozen) (1-10 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E; Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180430 475.00
Check your price
  • E-selectin Monoclonal Antibody for Western Blot, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122165
  • Size 100µg
Applications
ELISA (10 µg/ml), Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunohistochemistry (Frozen) (1-10 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122165 399.00
Check your price
  • E-Selectin/P-Selectin Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate FITC
  • Catalog # MA516428
  • Size 100ug
Applications
Flow Cytometry (Neat)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E; Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516428 325.00
Check your price
  • E-selectin Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # A15437
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15437 188.00
Check your price
  • E-selectin Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15436
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15436 198.00
Check your price
  • E-selectin Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A15438
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15438 289.00
Check your price