1 - 3 of 3 products displayed
  • CD151 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516443
  • Size 100ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516443 159.00
Check your price
  • CD151 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180804
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50 -1/100), Immunohistochemistry (Paraffin, Frozen) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180804 349.00
Check your price
  • CD151 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA181266
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181266 425.00
Check your price