1 - 30 of 30 products displayed
  • HisG Epitope Tag Monoclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # R94125
  • Size 50 µl
Applications
ELISA (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94125 226.00
Check your price
  • HisG Epitope Tag Monoclonal Antibody, AP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate AP
  • Catalog # R94225
  • Size 125 µl
Applications
Western Blot (1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94225 226.00
Check your price
  • HisG Epitope Tag Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # R94025
  • Size 50 µl
Applications
ELISA (Assay Dependent), Immunocytochemistry (2-4µg/ml), Immunofluorescence (2-4 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag, His G, 6xHis-Gly, 6xHis-Glycine
Catalog # Size Price (AUD) Your Price (AUD) Qty
R94025 184.00
Check your price
  • 6x-His Tag Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 710286
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1-3 ug/ml)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
710286 303.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1135
  • Size 100 ug
Applications
ELISA (25-500 ng/ml), Immunoprecipitation (2 ug), Western Blot (1:250-1:1000)
Alternate names:
HIS Tag, Histidine tag, Poly histidine
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 680
  • Catalog # MA121315D680
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D680 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 550
  • Catalog # MA121315D550
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D550 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 488
  • Catalog # MA121315D488
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D488 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight® 650
  • Catalog # MA121315D650
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315D650 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Biotin
  • Catalog # MA121315BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay Dependent), Western Blot (1:500-1000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315BTIN 159.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA121315HRP
  • Size 50 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315HRP 169.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1213151MG
  • Size 1 mg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:3000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1213151MG 950.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA121315
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:3000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315 159.00
Check your price
  • HA Tag Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1985
  • Size 100 µg
Applications
Western Blot (1:500-1:5000)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1985 249.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 647
  • Catalog # MA121315A647
  • Size 50 µl
Applications
Western Blot (1:500)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A647 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 555
  • Catalog # MA121315A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A555 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 488
  • Catalog # MA121315A488
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121315A488 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 647
  • Catalog # MA1135A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A647 219.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 555
  • Catalog # MA1135A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A555 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate Alexa Fluor® 488
  • Catalog # MA1135A488
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135A488 219.00
Check your price
  • 6x-His Tag Monoclonal Antibody, AP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate AP
  • Catalog # R93225
  • Size 125 µl
Applications
Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
R93225 230.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # R93125
  • Size 50 µl
Applications
Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
R93125 238.00
Check your price
  • 6x-His Tag Monoclonal Antibody for Western Blot, IF, ICC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # R93025
  • Size 50 µl
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunoprecipitation (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
R93025 197.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA1135HRP
  • Size 50 µl
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1135HRP 225.00
Check your price
  • 6x-His Tag Monoclonal Antibody, FITC conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Tag
  • Conjugate FITC
  • Catalog # R93325
  • Size 50 µl
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:1000-1:5000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
R93325 226.00
Check your price
  • 6x-His Tag Monoclonal Antibody, FITC conjugate for IF and Flow
  • Host Mouse
  • Target Species Tag
  • Conjugate FITC
  • Catalog # MA181891
  • Size 100 µg
Applications
Flow Cytometry (1/10), Immunofluorescence (Assay dependent)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181891 309.00
Check your price
  • 6x-His Tag Monoclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Mouse
  • Target Species Tag
  • Conjugate HRP
  • Catalog # MA180218
  • Size 100 µg
Applications
ELISA (1:20-1:1000), Western Blot (1/1000 -1/5,000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180218 449.00
Check your price
  • 6x-His Tag Polyclonal Antibody for Western Blot, ICC and ELISA
  • Host Chicken
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA19531
  • Size 200 ug
Applications
ELISA (1:1,000-1:5,000), Immunocytochemistry (1:400-1:2,000), Western Blot (1:1,000-1:5,000)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19531 275.00
Check your price
  • 6x-His Tag Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1983B
  • Size 200 µg
Applications
Western Blot (1:250)
Alternate names:
His Tag, Histidine Tag, Histidine Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1983B 249.00
Check your price
  • DYKDDDDK Tag Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA1984B
  • Size 200 µg
Applications
Immunofluorescence (1 ug/ml), Western Blot (1:500)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1984B 249.00
Check your price