1 - 2 of 2 products displayed
  • NT5C3A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124452
  • Size 50 µg
Applications
Western Blot (0.5-5 µg/ml)
Alternate names:
p36; Uridine 5'-monophosphate hydrolase 1; Pyrimidine 5'-nucleotidase 1; Cytosolic 5'-nucleotidase III; Cytosolic 5'-nucleotidase 3; 7-methylguanosine phosphate-specific 5'-nucleotidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124452 475.00
Check your price
  • NT5C3A Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540497
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
p36; Uridine 5'-monophosphate hydrolase 1; Pyrimidine 5'-nucleotidase 1; Cytosolic 5'-nucleotidase III; Cytosolic 5'-nucleotidase 3; 7-methylguanosine phosphate-specific 5'-nucleotidase; Lupin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540497 309.00
Check your price