1 - 30 of 30 products displayed
  • ADAM10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125166
  • Size 25 µg
Applications
Western Blot (1:500-1:2000)
Alternate names:
Mammalian disintegrin-metalloprotease; Kuzbanian protein homolog; CDw156
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125166 375.00
Check your price
  • ADAM9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517080
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma; Cellular disintegrin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517080 299.00
Check your price
  • ADAM15 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542864
  • Size 100 µl
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Metargidin; Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 15; Metalloprotease RGD disintegrin protein; CRII-7; AD56
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542864 309.00
Check your price
  • ADAM17 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527395
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527395 349.00
Check your price
  • ADAM10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112500
  • Size 100 µg
Applications
Western Blot (0.25-1 µg/ml)
Alternate names:
Mammalian disintegrin-metalloprotease; Kuzbanian protein homolog; CDw156
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112500 375.00
Check your price
  • ADAM17 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131074
  • Size 50 µg
Applications
peptide-ELISA (1:32,000), Western Blot (Assay dependent)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131074 349.00
Check your price
  • ADAM17 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519872
  • Size 100 ug
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519872 349.00
Check your price
  • ADAM17 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112517
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112517 325.00
Check your price
  • ADAM17 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA516551
  • Size 500 µL
Applications
Immunoprecipitation (10 µg/ml), Western Blot (5 µg/mL)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516551 399.00
Check your price
  • Phospho-CD156b / ADAM17 pThr735 Antibody Polyclonal (PA5-35619)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535619
  • Size
Applications
Western Blot (1:500-1:2000)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535619 339.00
Check your price
  • ADAM17 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131481
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
TNF-alpha-converting enzyme; TNF-alpha convertase; Snake venom-like protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131481 475.00
Check your price
  • ADAM12 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120301
  • Size 25 µg
Applications
Western Blot (1:1000-1:5000)
Alternate names:
Meltrin-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120301 375.00
Check your price
  • ADAM33 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518379
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
ADAM33; a disintegrin and metalloproteinase domain 33; HGNC:15478; DJ964F7.1; DKFZp434K0521; FLJ35308; FLJ36751; MGC71889; a disintegrin and metalloprotease 33; disintegrin and reprolysin metalloproteinase family protein; metalloprotease disintegrin; ADAM metallopeptidase domain 33; RP5-964F7.2; C20orf153; MGC149823
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518379 325.00
Check your price
  • ADAM20 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532652
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15-20 µg/ml)
Alternate names:
ADAM 20, a disintegrin and metalloproteinase domain 20, disintegrin and metalloproteinase domain-containing protein 20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532652 325.00
Check your price
  • ADAM20 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532651
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
ADAM 20, a disintegrin and metalloproteinase domain 20, disintegrin and metalloproteinase domain-containing protein 20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532651 325.00
Check your price
  • ADAM30 Antibody (Monoclonal, 3E5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521251
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
ADAM metallopeptidase domain 30, svph4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521251 309.00
Check your price
  • ADAM30 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA543393
  • Size 100 µl
Applications
Western Blot (5 µg/ml)
Alternate names:
ADAM metallopeptidase domain 30, svph4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543393 309.00
Check your price
  • ADAM33 Antibody (Monoclonal, 219515)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523936
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RP23-228H14.4, Adaml, ADAM 33, a disintegrin and metalloprotease domain 33, disintegrin and metalloproteinase domain-containing protein 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523936 299.00
Check your price
  • ADAM33 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547317
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
RP23-228H14.4, Adaml, ADAM 33, a disintegrin and metalloprotease domain 33, disintegrin and metalloproteinase domain-containing protein 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547317 325.00
Check your price
  • ADAM33 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA534614
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
RP23-228H14.4, Adaml, ADAM 33; a disintegrin and metalloprotease domain 33; disintegrin and metalloproteinase domain-containing protein 33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534614 325.00
Check your price
  • ADAM9 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547968
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma; Cellular disintegrin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547968 325.00
Check your price
  • ADAM9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514338
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma; Cellular disintegrin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514338 349.00
Check your price
  • ADAM9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525959
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma; Cellular disintegrin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525959 325.00
Check your price
  • ADAM22 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530639
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2; Metalloproteinase-disintegrin ADAM22-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530639 349.00
Check your price
  • ADAM9 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547970
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Myeloma cell metalloproteinase; Metalloprotease/disintegrin/cysteine-rich protein 9; Meltrin-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547970 325.00
Check your price
  • ADAM23 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530939
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530939 349.00
Check your price
  • ADAM23 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547678
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547678 325.00
Check your price
  • ADAM23 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127616
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Metalloproteinase-like, disintegrin-like, and cysteine-rich protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127616 399.00
Check your price
  • ADAMDEC1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544267
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
A disintegrin and metalloproteinase domain-like protein decysin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544267 309.00
Check your price
  • ADAM32 Antibody (Monoclonal, 539220)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524239
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
a disintegrin and metalloprotease domain 32, a disintegrin and metalloproteinase domain 32, ADAM 32, ADAM metallopeptidase domain 32, disintegrin and metalloproteinase domain-containing protein 32, FLJ26299, FLJ29004, metalloproteinase 12-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524239 299.00
Check your price