1 - 3 of 3 products displayed
  • AKAP9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 360300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein yotiao; Protein kinase A-anchoring protein 9; Protein hyperion; Centrosome- and Golgi-localized PKN-associated protein; AKAP 120-like protein; A-kinase anchor protein 450 kDa; A-kinase anchor protein 350 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
360300 304.00
Check your price
  • AKAP450 Antibody (Monoclonal, 17G10)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1803
  • Size 100 µg
Applications
Immunocytochemistry (4 µg/mL), Immunofluorescence (4 µg/mL), Immunohistochemistry (1:20), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay dependent), Western Blot (2 µg/mL)
Alternate names:
Protein yotiao; Protein kinase A-anchoring protein 9; Protein hyperion; Centrosome- and Golgi-localized PKN-associated protein; AKAP 120-like protein; A-kinase anchor protein 450 kDa; A-kinase anchor protein 350 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1803 349.00
Check your price
  • AKAP450 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518345
  • Size 100 ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Protein yotiao; Protein kinase A-anchoring protein 9; Protein hyperion; Centrosome- and Golgi-localized PKN-associated protein; AKAP 120-like protein; A-kinase anchor protein 450 kDa; A-kinase anchor protein 350 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518345 349.00
Check your price