1 - 2 of 2 products displayed
  • AA5 Virus VP3 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123359
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
HADV-5, AdV5, AV5, Adeno-associated virus 5, AAV-5 VP3, Adeno-associated 5 Virus VP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123359 375.00
Check your price
  • AA5 Virus VP3 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA522817
  • Size 100 ug
Applications
Western Blot (1:500-1:1000)
Alternate names:
HADV-5, AdV5, AV5, Adeno-associated virus 5, AAV-5 VP3, Adeno-associated 5 Virus VP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522817 349.00
Check your price