1 - 30 of 30 products displayed
  • Acetyl-CoA Carboxylase Antibody (Monoclonal, B.800.8)
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515025
  • Size 100 µL
Applications
Flow Cytometry (1:400), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
ACC-alpha; Acetyl-CoA carboxylase 265
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515025 275.00
Check your price
  • Acetyl-CoA Carboxylase Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517564
  • Size 100 µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
ACC-alpha; Acetyl-CoA carboxylase 265
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517564 275.00
Check your price
  • Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538079
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
ACC-alpha; Acetyl-CoA carboxylase 265
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538079 309.00
Check your price
  • Acetyl-CoA Carboxylase Antibody (Monoclonal, 143)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 437100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
ACC-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
437100 304.00
Check your price
  • Phospho-Acetyl-CoA Carboxylase (Ser79) Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517725
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:800), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
ACC-alpha; Acetyl-CoA carboxylase 265
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517725 309.00
Check your price
  • Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538107
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (Assay-Dependent)
Alternate names:
ACC-alpha; Acetyl-CoA carboxylase 265
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538107 309.00
Check your price
  • ACSS2 Antibody (Monoclonal, T.407.4)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514810
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Acyl-activating enzyme; Acyl-CoA synthetase short-chain family member 2; Acetyl-CoA synthetase; Acetate--CoA ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514810 275.00
Check your price
  • ACAT2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124413
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124413 399.00
Check your price
  • ACOT12 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531981
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
START domain-containing protein 15; Cytoplasmic acetyl-CoA hydrolase 1; Acyl-CoA thioester hydrolase 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531981 349.00
Check your price
  • ACSS2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526612
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Acyl-activating enzyme; Acyl-CoA synthetase short-chain family member 2; Acetyl-CoA synthetase; Acetate--CoA ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526612 349.00
Check your price
  • ACSS1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539002
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Acyl-CoA synthetase short-chain family member 1; Acetyl-CoA synthetase 2; Acetate--CoA ligase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539002 309.00
Check your price
  • ACOT12 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532152
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
START domain-containing protein 15; Cytoplasmic acetyl-CoA hydrolase 1; Acyl-CoA thioester hydrolase 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532152 349.00
Check your price
  • Phospho-AMPK alpha-1,2 (Thr172, Thr183) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536764
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536764 349.00
Check your price
  • Phospho-AMPK alpha-1,2 (Ser485, Ser491) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517543
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517543 309.00
Check your price
  • Phospho-AMPK alpha-1,2 (Thr172, Thr183) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536885
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536885 349.00
Check your price
  • Phospho-AMPK alpha-1 (Ser486) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536615
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Tau-protein kinase PRKAA1; Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536615 349.00
Check your price
  • AMPK alpha-1,2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536045
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536045 309.00
Check your price
  • AMPK alpha-1,2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536916
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536916 309.00
Check your price
  • AMPK alpha-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514026
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Tau-protein kinase PRKAA1; Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514026 349.00
Check your price
  • Phospho-AMPK alpha-1,2 (Thr172) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441150G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441150G 304.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529679
  • Size 100 µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529679 349.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517396
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517396 275.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514306
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514306 349.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514029
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514029 349.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525987
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525987 349.00
Check your price
  • ACAA1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529956
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Peroxisomal 3-oxoacyl-CoA thiolase A; Beta-ketothiolase A; Acetyl-CoA acyltransferase A; Peroxisomal 3-oxoacyl-CoA thiolase; Beta-ketothiolase; Acetyl-CoA acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529956 349.00
Check your price
  • ACAA1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529957
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Peroxisomal 3-oxoacyl-CoA thiolase; Beta-ketothiolase; Acetyl-CoA acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529957 349.00
Check your price
  • AMPK alpha-2 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519367
  • Size 100 ug
Applications
ELISA (1.5-3.0 ug/ml), Western Blot (0.3-1.0 ug/ml)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519367 349.00
Check your price
  • ACAT2 Antibody (Monoclonal, 4A5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518338
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518338 309.00
Check your price
  • ACAT2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540598
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540598 309.00
Check your price