1 - 24 of 24 products displayed
  • Actin Antibody (Monoclonal, mAbGEa)
  • Host Mouse
  • Target Species Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA1744
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Cytoplasmic beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1744 375.00
Check your price
  • Actin Antibody, Biotin (Monoclonal, ACTN05 (C4))
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • beta Actin Loading Control Antibody, DyLight® 488 (Monoclonal, BA3R)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 488 See Additional Formats Biotin DyLight 550 DyLight 650 DyLight 680 DyLight 800 4X PEG HRP Unconjugated
  • Catalog # MA515739D488
  • Size 50 µL
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D488 159.00
Check your price
  • beta Actin Loading Control Antibody (Monoclonal, BA3R)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated See Additional Formats Biotin DyLight 488 DyLight 550 DyLight 650 DyLight 680 DyLight 800 4X PEG HRP
  • Catalog # MA515739
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739 225.00
Check your price
  • ZO-1 Antibody
  • Host Rabbit
  • Target Species Dog, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 617300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (5-10µg/ml), Immunofluorescence (5-10µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Zonula occludens protein 1; Zona occludens protein 1; Tight junction protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
617300 362.00
Check your price
  • alpha Tubulin Antibody (Monoclonal, YL1/2)
  • Host Rat
  • Target Species Dog, Fruit fly, Human, Mouse, Plant, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # MA180017
  • Size 500 µg
Applications
ELISA (1/100-1/1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180017 399.00
Check your price
  • HSP70 Antibody (Monoclonal, 3A3)
  • Host Mouse
  • Target Species Amphibian, Arthropod, Avian, Fruit fly, Fish, Human, Mollusc, Mouse, Non-human primate, Plant, Pig, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA3006
  • Size 50 µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:100), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (2 µl), Western Blot (1:1000-1:5000)
Alternate names:
Hsp68; Heat shock 70 kDa protein 3; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3006 375.00
Check your price
  • ZO-2 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 711400
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Zonula occludens protein 2; Zona occludens protein 2; Tight junction protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
711400 303.00
Check your price
  • ZO-3 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 364100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunoprecipitation (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Zonula occludens protein 3; Zona occludens protein 3; Tight junction protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
364100 309.00
Check your price
  • GST Tag Antibody (Monoclonal, 1H14L28)
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 700775
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
Glutathione-S-Transferase, GST protein, GST Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
700775 304.00
Check your price
  • CaMKIV Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Plant, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA1542
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (1 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
CaM kinase-GR; Calspermin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1542 375.00
Check your price
  • Claudin 16 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 345400
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Paracellin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
345400 305.00
Check your price
  • V5 Tag Antibody (Monoclonal, 2F11F7)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 377500
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (1.0 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
377500 265.00
Check your price
  • Histone H3 Antibody (Monoclonal, 865R2)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # AHO1432
  • Size 100 µg
Applications
Western Blot (0.5-2 µg/mL)
Alternate names:
Histone H3/l; Histone H3/k; Histone H3/j; Histone H3/i; Histone H3/h; Histone H3/f; Histone H3/d; Histone H3/c; Histone H3/b; Histone H3/a
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1432 306.00
Check your price
  • Claudin 7 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 349100
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CEPTRL2, CLDN-7, CPETRL2, Hs.84359, claudin-1, claudin-7, clostridium perfringens enterotoxin receptor-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
349100 308.00
Check your price
  • BMAL1 Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1523
  • Size 200 µL
Applications
Western Blot (1:200)
Alternate names:
Brain and muscle ARNT-like 1; Arnt3; bHLH-PAS protein JAP3; PAS domain-containing protein 3; Member of PAS protein 3; Class E basic helix-loop-helix protein 5; Basic-helix-loop-helix-PAS protein MOP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1523 375.00
Check your price
  • Bi-1 Antibody (Monoclonal, 20F248)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141678
  • Size 0.1 ml
Applications
Western Blot (1:500-1:1000)
Alternate names:
Transmembrane BAX inhibitor motif-containing protein 6; Testis-enhanced gene transcript protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141678 309.00
Check your price
  • PER1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1524
  • Size 200 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin, Frozen) (1:50-1:500), Western Blot (1:100-1:1000)
Alternate names:
Circadian pacemaker protein Rigui; Circadian clock protein PERIOD 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1524 375.00
Check your price
  • CLOCK Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1520
  • Size 200 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin, Frozen) (1:100-1:1000), Western Blot (1:200)
Alternate names:
Class E basic helix-loop-helix protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1520 375.00
Check your price
  • 14-3-3 eta Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529774
  • Size 100 µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529774 349.00
Check your price
  • 14-3-3 eta Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528113
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528113 349.00
Check your price
  • 14-3-3 eta Antibody
  • Host Rabbit
  • Target Species Dog, Human, Rabbit, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA546811
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Protein AS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546811 309.00
Check your price
  • CRY1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1527
  • Size 100 µg
Applications
Western Blot (1:2,000)
Alternate names:
Cry1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1527 349.00
Check your price
  • ATP Synthase beta Antibody (Monoclonal, 3D5AB1)
  • Host Mouse
  • Target Species Bovine, C. elegans, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # A21351
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Western Blot (0.5 ug/ml)
Alternate names:
ATPMB, ATPSB, MGC5231
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21351 330.00
Check your price