1 - 30 of 30 products displayed
  • ASK Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 407300
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
DBF4-type zinc finger-containing protein 1; Chiffon homolog A; Activator of S phase kinase; MuDBF4
Catalog # Size Price (AUD) Your Price (AUD) Qty
407300 309.00
Check your price
  • DBF4B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535871
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000)
Alternate names:
Dbf4-related factor 1; Chiffon homolog B; Activator of S phase kinase-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535871 309.00
Check your price
  • Phospho-p70 S6 Kinase (Thr389) Antibody (Oligoclonal, B2HCLC)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 710095
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710095 309.00
Check your price
  • p70 S6 Kinase Antibody (Oligoclonal, 15HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710285
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:100-1:1000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710285 304.00
Check your price
  • p70 S6 Kinase Antibody (Monoclonal, 15H5L2)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701261
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701261 309.00
Check your price
  • DNA-PK Antibody (Cocktail, 18-2 + 25-4 + 42-psc)
  • Host Mouse
  • Target Species Chicken, Hamster, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA513404
  • Size 500 µL
Applications
Gel Shift (1mg/mL), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Immunoprecipitation (2 µg/mg protein lysate), Western Blot (1-2 µg/ml)
Alternate names:
p460; DNPK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513404 349.00
Check your price
  • Phospho-p70 S6 Kinase (Thr229) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44918G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Western Blot (Assay Dependent)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44918G 309.00
Check your price
  • Phospho-p70 S6 Kinase (Thr421, Ser424) Antibody (Oligoclonal, B12HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710108
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
p70 ribosomal S6 kinase alpha; Serine/threonine-protein kinase 14A; Ribosomal protein S6 kinase I; 70 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710108 309.00
Check your price
  • Caspase 9 (active) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA126434
  • Size 50 µg
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 ug/ml), Immunoprecipitation (10-20 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
ICE-like apoptotic protease 6; Apoptotic protease-activating factor 3; Apoptotic protease Mch-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126434 399.00
Check your price
  • Caspase 3 (active/pro) Antibody (Monoclonal, 31A1067)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191637
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (2-3 µg/ml)
Alternate names:
SREBP cleavage activity 1; Protein Yama; Cysteine protease CPP32; Apopain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191637 425.00
Check your price
  • active Caspase 9 Antibody Polyclonal (PA1-26435)
  • Host Rabbit
  • Target Species Rat
  • Conjugate
  • Catalog # PA126435
  • Size 50µg
Applications
Immunocytochemistry (10-20 µg/ml), Immunofluorescence (10-20 ug/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
RP11-265F14.3, APAF-3, APAF3, CASPASE-9c, ICE-LAP6, MCH6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126435 369.00
Check your price
  • SKP1 Antibody (Monoclonal, 4E11)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515928
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p19skp1; p19A; S-phase kinase-associated protein 1A; Cyclin-A/CDK2-associated protein p19; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515928 349.00
Check your price
  • SKP1 Antibody (Monoclonal, 1H9)
  • Host Mouse
  • Target Species Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA517174
  • Size 100 µL
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
p19skp1; p19A; S-phase kinase-associated protein 1A; Cyclin-A/CDK2-associated protein p19; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517174 309.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517468
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
p19skp1; p19A; S-phase kinase-associated protein 1A; Cyclin-A/CDK2-associated protein p19; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517468 275.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532584
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532584 349.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511772
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511772 349.00
Check your price
  • SKP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 511900
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5 ug), Western Blot (1-2 ug/ml)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
511900 304.00
Check your price
  • SKP2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517009
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:1000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45; F-box/WD-40 protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517009 275.00
Check your price
  • SKP2 Antibody (Monoclonal, 6G9D10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515470
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515470 349.00
Check your price
  • SKP2 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517497
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:1000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517497 275.00
Check your price
  • SKP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535331
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535331 349.00
Check your price
  • SKP2 Antibody (Monoclonal, SKP2-8D9)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 323300
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5 ug), Western Blot (1-2 ug/ml)
Alternate names:
p45skp2; F-box/LRR-repeat protein 1; F-box protein Skp2; Cyclin-A/CDK2-associated protein p45
Catalog # Size Price (AUD) Your Price (AUD) Qty
323300 304.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528489
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528489 349.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529424
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529424 349.00
Check your price
  • SKP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519442
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519442 349.00
Check your price
  • SKP1 Antibody (Monoclonal, 1C10F4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 323800
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5-10 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
p19skp1; Transcription elongation factor B polypeptide 1-like; SIII; RNA polymerase II elongation factor-like protein; Organ of Corti protein II; Organ of Corti protein 2; Cyclin-A/CDK2-associated protein p19
Catalog # Size Price (AUD) Your Price (AUD) Qty
323800 304.00
Check your price
  • BMPR1A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 386000
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase receptor R5; BMP-2/BMP-4 receptor; Activin receptor-like kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
386000 305.00
Check your price
  • GRK2 Antibody (Monoclonal, 5D5)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA12013
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay dependent), Western Blot (3-5 ug/ml)
Alternate names:
G-protein-coupled receptor kinase 2; G-protein coupled receptor kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12013 275.00
Check your price
  • Aurora A Antibody (Monoclonal, 35C1)
  • Host Mouse
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # 458900
  • Size 100 µg
Applications
Immunocytochemistry (1-2µg/mL), Immunofluorescence (1-2µg/mL), Immunoprecipitation (5 ug), Western Blot (1-3 µg/ml)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase 15; Aurora/IPL1-related kinase 1; Aurora 2; Serine/threonine-protein kinase Ayk1; Ipl1- and aurora-related kinase 1; Aurora family kinase 1; Breast tumor-amplified kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
458900 304.00
Check your price
  • Phospho-ERK1/ERK2 (Thr202, Tyr204) Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 368800
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:500), Western Blot (1-3 µg/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
368800 308.00
Check your price