1 - 30 of 30 products displayed
  • RhoA Antibody (Monoclonal, 1B8-1C7)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1134
  • Size 100 ug
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (1:500-1:2000)
Alternate names:
Rho cDNA clone 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1134 309.00
Check your price
  • RhoA Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00266W
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Rho cDNA clone 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00266W 309.00
Check your price
  • RhoA Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00298G
  • Size 500 µg
Applications
Immunohistochemistry (10-50 µg/ml), Western Blot (10-50 µg/ml)
Alternate names:
Rho cDNA clone 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00298G 309.00
Check your price
  • RhoA/RhoC Antibody (Monoclonal, 1B3-4A10)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1123
  • Size 100 µg
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Immunoprecipitation (2 µg), Western Blot (1:500-1:1500)
Alternate names:
Silica-induced gene 61 protein; Rho cDNA clone 12; Rho cDNA clone 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1123 309.00
Check your price
  • RhoA/RhoB/RhoC Antibody (Monoclonal, 1A11-4G10)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1011
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:1500)
Alternate names:
Rho cDNA clone 6; Silica-induced gene 61 protein; Rho cDNA clone 12; Rho cDNA clone 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1011 309.00
Check your price
  • Rho Pan Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSC00025W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Rho cDNA clone 6; Silica-induced gene 61 protein; Ras-like protein TC10; Rho cDNA clone 12; Rho cDNA clone 9; Rho in filopodia; Rho family GTPase Rif; Ras-like protein family member 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSC00025W 309.00
Check your price
  • Rho Pan Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSR00297G
  • Size 500 µg
Applications
Immunohistochemistry (10-50 µg/ml), Western Blot (10-50 µg/ml)
Alternate names:
Rho cDNA clone 6; Silica-induced gene 61 protein; Ras-like protein TC10; Rho cDNA clone 12; Rho cDNA clone 9; Rho in filopodia; Rho family GTPase Rif; Ras-like protein family member 7A
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00297G 309.00
Check your price
  • Phospho-RhoA pSer188 Antibody Polyclonal (PA5-35486)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535486
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Rho cDNA clone 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535486 339.00
Check your price
  • Phospho-RACGAP1 (Ser387) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538575
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Male germ cell RacGap; Protein CYK4 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538575 309.00
Check your price
  • ROCK1 (Cleaved) Antibody (Monoclonal, 154C1465)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA141031
  • Size 100 µg
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1-2 µg/ml)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35; p150 RhoA-binding kinase ROK beta; Liver regeneration-related protein LRRG199; cAMP-dependent protein kinase ROCK-I; HEBM1; Corneal epithelial Rho-associated-Ser/Thr kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141031 375.00
Check your price
  • ARHGAP10 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524188
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Rho-type GTPase-activating protein 10; Graf-related protein 2; GTPase regulator associated with focal adhesion kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524188 349.00
Check your price
  • ARHGAP17 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524250
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
RhoGAP interacting with CIP4 homologs protein 1; Rho-type GTPase-activating protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524250 349.00
Check your price
  • GRAF Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518213
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518213 349.00
Check your price
  • ARHGAP19 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526812
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Rho-type GTPase-activating protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526812 349.00
Check your price
  • ROCK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521130
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35; p150 RhoA-binding kinase ROK beta; Liver regeneration-related protein LRRG199
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521130 349.00
Check your price
  • Phospho-ROCK2 (Tyr722) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537774
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
p164 ROCK-2; Rho-associated, coiled-coil-containing protein kinase II; Rho-associated, coiled-coil-containing protein kinase 2; Rho kinase 2; p150 ROK-alpha; RhoA-binding kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537774 309.00
Check your price
  • ROCK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521131
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
p164 ROCK-2; Rho-associated, coiled-coil-containing protein kinase II; Rho-associated, coiled-coil-containing protein kinase 2; Rho kinase 2; p150 ROK-alpha; RhoA-binding kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521131 349.00
Check your price
  • ROCK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528774
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p164 ROCK-2; Rho-associated, coiled-coil-containing protein kinase II; Rho-associated, coiled-coil-containing protein kinase 2; Rho kinase 2; p150 ROK-alpha; RhoA-binding kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528774 349.00
Check your price
  • GEFT Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544047
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT; Rac/Cdc42/Rho exchange factor GEFT; Guanine nucleotide exchange factor GEFT; p63RhoGEF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544047 309.00
Check your price
  • GEFT Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531827
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p63RhoGEF; RhoA/Rac/Cdc42 guanine nucleotide exchange factor GEFT; Rac/Cdc42/Rho exchange factor GEFT; Guanine nucleotide exchange factor GEFT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531827 349.00
Check your price
  • GRAF Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530967
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530967 349.00
Check your price
  • GRAF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528703
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528703 349.00
Check your price
  • GRAF Antibody
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131264
  • Size 100 µg
Applications
ELISA (Assay-dependent), Western Blot (0.3-1 µg/ml)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131264 375.00
Check your price
  • ARHGEF11 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523162
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.1-0.5 µg/ml)
Alternate names:
PDZ-RhoGEF; RhoGEF glutamate transport modulator GTRAP48
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523162 349.00
Check your price
  • ARHGAP19 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA545144
  • Size 100 µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Rho-type GTPase-activating protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545144 309.00
Check your price
  • ROCK1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA522262
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35; p150 RhoA-binding kinase ROK beta; Liver regeneration-related protein LRRG199
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522262 349.00
Check your price
  • LAMTOR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523086
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Immunohistochemistry (Paraffin) (1:25-1:100), Western Blot (1:1000)
Alternate names:
p27Kip1-releasing factor from RhoA; Protein associated with DRMs and endosomes; Lipid raft adaptor protein p18; Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523086 349.00
Check your price
  • MRI1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545103
  • Size 100 µl
Applications
Western Blot (1 µg/ml)
Alternate names:
Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein; S-methyl-5-thioribose-1-phosphate isomerase; Mediator of RhoA-dependent invasion
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545103 309.00
Check your price
  • DAAM1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523222
  • Size 100 ug
Applications
Western Blot (4.0-6 µg/ml)
Alternate names:
disheveled-associated activator of morphogenesis 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523222 349.00
Check your price
  • ROCK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFPA0048
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35; p150 RhoA-binding kinase ROK beta; Liver regeneration-related protein LRRG199
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0048 309.00
Check your price