1 - 30 of 30 products displayed
  • TGF beta-2 Antibody (Oligoclonal, 12HCLC)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710276
  • Size 100 µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Polyergin; Glioblastoma-derived T-cell suppressor factor; Cetermin; BSC-1 cell growth inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710276 309.00
Check your price
  • TGF beta Receptor I Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate
  • Catalog # PA527785
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527785 349.00
Check your price
  • TGFBR2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536115
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transforming growth factor-beta receptor type II; TGF-beta type II receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536115 309.00
Check your price
  • TGF beta-1,2,3 Antibody (Monoclonal, 1D11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523795
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Paraffin) (8-25 µg/ml), Inhibition Assays (Assay-Dependent), Neutralization (0.25-1.25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523795 375.00
Check your price
  • pro-TGF beta Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129020
  • Size 100 µg
Applications
Immunohistochemistry (1:200-1:500), Western Blot (1:2000)
Alternate names:
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129020 309.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, E8-4D12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1116
  • Size 200 ug
Applications
ELISA (1ug/ml), Western Blot (1ug/ml)
Alternate names:
TGFB, DPD1, TGF beta-1, LAP, TGF beta, Latency-Associated Peptide, TGF beta-1 protein, Transforming Growth Factor beta-1, TGF-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1116 309.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, B11-4C3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1169
  • Size 200 ug
Applications
ELISA (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, DPD1, TGF beta-1, LAP, TGF beta, Latency-Associated Peptide, TGF beta-1 protein, Transforming Growth Factor beta-1, TGF-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1169 309.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, 9016)
  • Host Mouse
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523702
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (Assay-Dependent), Neutralization (0.3-1.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523702 375.00
Check your price
  • TGF beta-3 Antibody
  • Host Sheep
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # PA112642
  • Size 250 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
TGF-beta3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112642 325.00
Check your price
  • TGF beta-3 Antibody (Monoclonal, 20724)
  • Host Mouse
  • Target Species Human, Many, Mouse
  • Conjugate Unconjugated
  • Catalog # MA523812
  • Size 100 µg
Applications
Immunohistochemistry (8-25 µg/ml), Neutralization (0.1-0.3 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TGFb3, ARVD, TGF-beta3, TGF-beta-3, transforming growth factor beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523812 275.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, F.888.7)
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA515065
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515065 275.00
Check your price
  • eIF3i Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527582
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
eIF3 p36; eIF-3-beta; TGF-beta receptor-interacting protein 1; Eukaryotic translation initiation factor 3 subunit 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527582 349.00
Check your price
  • TGF beta-1 Antibody
  • Host Chicken
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA19574
  • Size 100 µg
Applications
ELISA (1:2000-1:5000), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (1:2000-1:5000)
Alternate names:
CED, DPD1, TGFB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19574 375.00
Check your price
  • TGFBR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514959
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD; ESK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514959 349.00
Check your price
  • TGFBR2 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547719
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (2 µg/ml)
Alternate names:
Transforming growth factor-beta receptor type II; TGF-beta type II receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547719 375.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, TB21)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518023
  • Size 100 µL
Applications
ELISA (Assay-dependent), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (5-20 ng/mL)
Alternate names:
CED; DPD1; TGFB; TGFbeta; TGFB1; Latency-associated peptide; LAP; Transforming growth factor beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518023 425.00
Check your price
  • TGFBR3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521994
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Transforming growth factor beta receptor III; Betaglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521994 349.00
Check your price
  • TGFBR3 Antibody (Monoclonal, D11G10)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A16694
  • Size 100 µL
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Transforming growth factor beta receptor III; Betaglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16694 309.00
Check your price
  • TGIF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521900
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
TGF-beta-induced transcription factor 2; 5'-TG-3'-interacting factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521900 349.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, 2C5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126058
  • Size 250 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126058 425.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, TB21)
  • Host Mouse
  • Target Species Bovine, Human, Mustelid, Mink, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516949
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta,TGF-beta 1 protein; TGF-beta-1; latency-associated peptide; transforming growth factor beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516949 375.00
Check your price
  • TGF beta-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129032
  • Size 100 µg
Applications
Immunohistochemistry (1:200-1:500), Western Blot (1:2000)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129032 309.00
Check your price
  • eIF3i Antibody (Monoclonal, 3B10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520471
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
eIF3 p36; eIF-3-beta; TGF-beta receptor-interacting protein 1; Eukaryotic translation initiation factor 3 subunit 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520471 309.00
Check your price
  • TGFBR1 Antibody (Monoclonal, 234R32)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # AHO1552
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO1552 309.00
Check your price
  • TGF beta-1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1197
  • Size 100 ug
Applications
Western Blot (1:250-1:1000)
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1197 309.00
Check your price
  • TGFBR1 Antibody
  • Host Rabbit
  • Target Species Human, Mink, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA517723
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Transforming growth factor-beta receptor type I; TGF-beta type I receptor; Serine/threonine-protein kinase receptor R4; Activin receptor-like kinase 5; Activin A receptor type II-like protein kinase of 53kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517723 275.00
Check your price
  • TGFBR3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517529
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Transforming growth factor beta receptor III; Betaglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517529 275.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, 4F9C10,1B11B7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515289
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
CED; DPD1; TGFB; TGFbeta; TGFB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515289 349.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, TB21)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121595
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (Assay dependent), Neutralization (Assay dependent), Western Blot (5-20 ng/mL)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121595 375.00
Check your price
  • TGF beta-1 Antibody (Monoclonal, 2E6)
  • Host Mouse
  • Target Species Human, Sheep
  • Conjugate Unconjugated
  • Catalog # MA121479
  • Size 100 µg
Applications
ELISA (Assay dependent), Neutralization (Assay-dependent), Western Blot (1-10 ng/ml)
Alternate names:
CED, DPD1, LAP, TGFB, TGFbeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121479 375.00
Check your price