1 - 7 of 7 products displayed
  • ACVR2A Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522325
  • Size 100 µL
Applications
Western Blot (1:5000-1:20,000)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522325 349.00
Check your price
  • ACVR2A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120174
  • Size 25 µg
Applications
ELISA (0.5 ug/ml), Immunohistochemistry (Paraffin) (15 ug/ml), Neutralization (1-3 ug/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120174 349.00
Check your price
  • ACVR2A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547484
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547484 349.00
Check your price
  • ACVR2A Antibody (Monoclonal, 149/1)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA191751
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191751 375.00
Check your price
  • ACVR2A Antibody (Monoclonal, 149/1)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA191752
  • Size 500 µL
Applications
Immunoprecipitation (1:400), Western Blot (1:200)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191752 349.00
Check your price
  • ACVR2B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120175
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Neutralization (1-3 µg/ml), Western Blot (0.1-0.2 ug/ml)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120175 349.00
Check your price
  • ACVR2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513886
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513886 349.00
Check your price