1 - 17 of 17 products displayed
  • ACVR2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528231
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528231 349.00
Check your price
  • ACVR2B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120175
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Neutralization (1-3 µg/ml), Western Blot (0.1-0.2 ug/ml)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120175 349.00
Check your price
  • ACVR2B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547005
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547005 375.00
Check your price
  • ACVR2B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513888
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Activin receptor type IIB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513888 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513881
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I; TSK-7L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513881 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513882
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I; TSK-7L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513882 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514920
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I; TSK-7L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514920 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513880
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513880 349.00
Check your price
  • ACVR2A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA120174
  • Size 25 µg
Applications
ELISA (0.5 ug/ml), Immunohistochemistry (Paraffin) (15 ug/ml), Neutralization (1-3 ug/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120174 349.00
Check your price
  • ACVR1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514911
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase receptor R2; Activin receptor-like kinase 4; Activin receptor type IB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514911 349.00
Check your price
  • ACVR1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514912
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Serine/threonine-protein kinase receptor R2; Activin receptor-like kinase 4; Activin receptor type IB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514912 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA528662
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528662 349.00
Check your price
  • ACVR1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529219
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serine/threonine-protein kinase receptor R2; Activin receptor-like kinase 4; Activin receptor type IB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529219 349.00
Check your price
  • ACVR1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527806
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serine/threonine-protein kinase receptor R2; Activin receptor-like kinase 4; Activin receptor type IB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527806 349.00
Check your price
  • ACVR1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141185
  • Size 100 µg
Applications
Western Blot (3-5 µg/ml)
Alternate names:
TGF-B superfamily receptor type I; Serine/threonine-protein kinase receptor R1; Activin receptor-like kinase 2; Activin receptor type I; TSK-7L
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141185 349.00
Check your price
  • ACVR2A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513886
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Activin receptor type IIA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513886 349.00
Check your price
  • ACVR1B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA128007
  • Size 25 µg
Applications
ELISA (0.5-1 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
Serine/threonine-protein kinase receptor R2; Activin receptor-like kinase 4; Activin receptor type IB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128007 349.00
Check your price