1 - 19 of 19 products displayed
  • SLC27A6 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534544
  • Size 100 ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Very long-chain acyl-CoA synthetase homolog 1; Solute carrier family 27 member 6; Fatty-acid-coenzyme A ligase, very long-chain 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534544 349.00
Check your price
  • Human SLC27A6 Synthetic Peptide
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP1587
  • Size 50 ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP1587 85.75
Check your price
  • SLC27A2 Antibody (Monoclonal, 466106)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524186
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Very long-chain-fatty-acid-CoA ligase; THCA-CoA ligase; Solute carrier family 27 member 2; Long-chain-fatty-acid--CoA ligase; Fatty-acid-coenzyme A ligase, very long-chain 1; Fatty acid transport protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524186 275.00
Check your price
  • SLC27A2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530420
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Very long-chain-fatty-acid-CoA ligase; THCA-CoA ligase; Solute carrier family 27 member 2; Long-chain-fatty-acid--CoA ligase; Fatty-acid-coenzyme A ligase, very long-chain 1; Fatty acid transport protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530420 349.00
Check your price
  • SLC27A2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542429
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Very long-chain-fatty-acid-CoA ligase; THCA-CoA ligase; Solute carrier family 27 member 2; Long-chain-fatty-acid--CoA ligase; Fatty-acid-coenzyme A ligase, very long-chain 1; Fatty acid transport protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542429 309.00
Check your price
  • ACSL3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512222
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Long-chain acyl-CoA synthetase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512222 349.00
Check your price
  • ACSL4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518099
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Long-chain acyl-CoA synthetase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518099 349.00
Check your price
  • ACSL4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512225
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Long-chain acyl-CoA synthetase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512225 349.00
Check your price
  • ACSL4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527137
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Long-chain acyl-CoA synthetase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527137 349.00
Check your price
  • ACSL4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530026
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Long-chain acyl-CoA synthetase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530026 349.00
Check your price
  • ACSL3 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542883
  • Size 100 µl
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (1:200), Western Blot (0.2-1 µg/ml)
Alternate names:
Long-chain acyl-CoA synthetase 3; Brain acyl-CoA synthetase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542883 309.00
Check your price
  • ACSL3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529507
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Long-chain acyl-CoA synthetase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529507 349.00
Check your price
  • SLC27A5 Antibody (Monoclonal, 4B11C10)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA517175
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Very long-chain acyl-CoA synthetase-related protein; Solute carrier family 27 member 5; Fatty acid transport protein 5; Cholate--CoA ligase; Bile acid-CoA ligase; Very long-chain acyl-CoA synthetase homolog 2; Fatty-acid-coenzyme A ligase, very long-chain 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517175 309.00
Check your price
  • SLC27A5 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542028
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Very long-chain acyl-CoA synthetase-related protein; Solute carrier family 27 member 5; Fatty acid transport protein 5; Cholate--CoA ligase; Bile acid-CoA ligase; Very long-chain acyl-CoA synthetase homolog 2; Fatty-acid-coenzyme A ligase, very long-chain 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542028 309.00
Check your price
  • SLC27A5 Antibody (Monoclonal, 390917)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524125
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Very long-chain acyl-CoA synthetase-related protein; Very long-chain acyl-CoA synthetase homolog 2; Solute carrier family 27 member 5; Fatty-acid-coenzyme A ligase, very long-chain 3; Fatty acid transport protein 5; Cholate--CoA ligase; Bile acid-CoA ligase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524125 275.00
Check your price
  • SLC27A3 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542490
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Very long-chain acyl-CoA synthetase homolog 3; Solute carrier family 27 member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542490 309.00
Check your price
  • ACSBG2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526853
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Bubblegum-related protein; Acyl-CoA synthetase bubblegum family member 2; PRTD-NY3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526853 349.00
Check your price
  • ACSF3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525803
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
acyl-CoA synthetase family member 3, mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525803 349.00
Check your price
  • SLC27A4 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA542446
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 27 member 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542446 309.00
Check your price