1 - 7 of 7 products displayed
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532658
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (14 µg/ml)
Alternate names:
Aggrecanase-2; ADMP-2; A disintegrin and metalloproteinase with thrombospondin motifs 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532658 349.00
Check your price
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA514350
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Implantin; Aggrecanase-2; ADMP-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514350 349.00
Check your price
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532142
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Aggrecanase-2; ADMP-2; A disintegrin and metalloproteinase with thrombospondin motifs 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532142 349.00
Check your price
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA11742
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:100-1:1000)
Alternate names:
Aggrecanase-2; ADMP-2; A disintegrin and metalloproteinase with thrombospondin motifs 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11742 275.00
Check your price
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA11751A
  • Size 100 µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Aggrecanase-2; ADMP-2; A disintegrin and metalloproteinase with thrombospondin motifs 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11751A 309.00
Check your price
  • ADAMTS5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527165
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Aggrecanase-2; ADMP-2; A disintegrin and metalloproteinase with thrombospondin motifs 11; Implantin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527165 349.00
Check your price
  • Human ADAMTS5 Recombinant Protein
  • Host Insect cells
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77529
  • Size 5 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77529 524.00
Check your price