1 - 2 of 2 products displayed
  • AP1S2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522018
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Sigma1B-adaptin; Sigma-adaptin 1B; Sigma 1B subunit of AP-1 clathrin; Golgi adaptor HA1/AP1 adaptin sigma-1B subunit; Clathrin assembly protein complex 1 sigma-1B small chain; Adaptor-related protein complex 1 subunit sigma-1B; Adaptor protein complex AP-1 subunit sigma-1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522018 349.00
Check your price
  • AP1S2 Antibody (Monoclonal, 3B9-G5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520556
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Sigma1B-adaptin; Sigma-adaptin 1B; Sigma 1B subunit of AP-1 clathrin; Golgi adaptor HA1/AP1 adaptin sigma-1B subunit; Clathrin assembly protein complex 1 sigma-1B small chain; Adaptor-related protein complex 1 subunit sigma-1B; Adaptor protein complex AP-1 subunit sigma-1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520556 309.00
Check your price