1 - 30 of 30 products displayed
  • TMEM161A Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA543020
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Adaptive response to oxidative stress protein 29
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543020 309.00
Check your price
  • SQSTM1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA118271
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:500-1:5000)
Alternate names:
Ubiquitin-binding protein p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118271 399.00
Check your price
  • SQSTM1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534781
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Ubiquitin-binding protein p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534781 349.00
Check your price
  • SQSTM1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520839
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (0.2-2 ug/ml)
Alternate names:
Ubiquitin-binding protein p62; Protein kinase C-zeta-interacting protein; STONE14; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520839 349.00
Check your price
  • SQSTM1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512092
  • Size 400 µL
Applications
Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Ubiquitin-binding protein p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512092 349.00
Check your price
  • SQSTM1 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSS00006W
  • Size 150 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:200-1:2000), Western Blot (1:200-1:2000)
Alternate names:
Ubiquitin-binding protein p62; Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; EBI3-associated protein of 60 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSS00006W 309.00
Check your price
  • HIF1A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1184
  • Size 100 µg
Applications
ELISA (1:1000), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1184 349.00
Check your price
  • Nurr1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14519
  • Size 100 µg
Applications
Immunofluorescence (1:100-1:200), Immunohistochemistry (1:100-1:200), Western Blot (1:1000-1:2000)
Alternate names:
Orphan nuclear receptor NURR1; NUR-related factor 1; Transcriptionally-inducible nuclear receptor; Immediate-early response protein NOT; SL-322; Regenerating liver nuclear receptor 1; Nuclear orphan receptor HZF-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14519 349.00
Check your price
  • SHC Antibody (Monoclonal, 47F4)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0201
  • Size 100 µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 ug), Western Blot (0.1 ug/ml)
Alternate names:
Src homology 2 domain-containing-transforming protein C1; SHC-transforming protein A; SHC-transforming protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0201 309.00
Check your price
  • SIRT1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523063
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1:150), Western Blot (1:1000)
Alternate names:
SIR2-like protein 1; Regulatory protein SIR2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523063 349.00
Check your price
  • Nurr1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA522830
  • Size 100 ug
Applications
Immunofluorescence (Assay-dependent), Immunohistochemistry (1:100-1:200), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2000)
Alternate names:
Transcriptionally-inducible nuclear receptor; Orphan nuclear receptor NURR1; Immediate-early response protein NOT; SL-322; Regenerating liver nuclear receptor 1; Nuclear orphan receptor HZF-3; NUR-related factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522830 349.00
Check your price
  • PYK2 Antibody (Monoclonal, 9H12L1)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700183
  • Size 100 µg
Applications
ChIP assay (1 ul), Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (4-6 ug/ml), Western Blot (1-3µg/ml)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
700183 303.00
Check your price
  • ARC Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529861
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nucleolar protein of 30 kDa; Muscle-enriched cytoplasmic protein; Apoptosis repressor with CARD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529861 349.00
Check your price
  • TRAF2 Antibody (Monoclonal, 33A1293)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141028
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (2 µg/ml)
Alternate names:
Tumor necrosis factor type 2 receptor-associated protein 3; E3 ubiquitin-protein ligase TRAF2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141028 309.00
Check your price
  • PINK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523072
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:1000), Immunofluorescence (1:50-1:1000), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
PTEN-induced putative kinase protein 1; BRPK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523072 349.00
Check your price
  • ARC Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116860
  • Size 100 µg
Applications
Western Blot (1:100-1:2000)
Alternate names:
Nucleolar protein of 30 kDa; Muscle-enriched cytoplasmic protein; Apoptosis repressor with CARD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116860 309.00
Check your price
  • Phospho-JAK2 (Tyr221) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # A14017
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
Janus kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14017 309.00
Check your price
  • HIF1A Antibody (Oligoclonal, 16HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710059
  • Size 100 µg
Applications
ChIP assay (3µg), ELISA (Assay-Dependent), Immunocytochemistry (1µg/ml), Immunofluorescence (1µg/ml), Western Blot (0.5-1µg/ml)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710059 305.00
Check your price
  • HIF1A Antibody (Monoclonal, 16H4L13)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700505
  • Size 100 µg
Applications
ChIP assay (1 ul), ELISA (Assay-dependent), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:5000)
Alternate names:
PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1; ARNT-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
700505 304.00
Check your price
  • CaMKII beta/gamma/delta Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536562
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG, Calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II subunit delta, CaMK-II subunit delta, CAMK2D, CAMKD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536562 309.00
Check your price
  • CaMKII beta/gamma Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536195
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536195 309.00
Check your price
  • SIRT1 Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517074
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
SIR2alpha; SIR2-like protein 1; Regulatory protein SIR2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517074 275.00
Check your price
  • Phospho-SIRT1 (Ser47) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517391
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:25), Western Blot (1:2000)
Alternate names:
SIR2-like protein 1; Regulatory protein SIR2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517391 309.00
Check your price
  • SIRT1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517393
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
SIR2-like protein 1; Regulatory protein SIR2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517393 275.00
Check your price
  • SIRT1 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA517232
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:25), Western Blot (1:1000)
Alternate names:
SIR2-like protein 1; Regulatory protein SIR2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517232 275.00
Check your price
  • Phospho-CaMKII beta/gamma/delta (Thr287) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537833
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, Calcium/calmodulin-dependent protein kinase type II subunit delta, Ca, CAMKD, CaM kinase II delta subunit, CaM-kinase II delta chain, CaMK-II delta subunit, caM kinase II subunit delta, caMK-II subunit delta, calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CAMK, CAMK-II, CAMKG, caMK-II subunit gamma, calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537833 309.00
Check your price
  • Phospho-CaMKII beta/gamma/delta (Thr287) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539731
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539731 309.00
Check your price
  • CaMKII beta/gamma/delta Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539732
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II subunit beta, CaMK-II subunit beta, CAMK2B, CAM2, CAMK2, CAMKB, Calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CaMK-II subunit gamma, CAMK2G, CAMK, CAMK-II, CAMKG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539732 309.00
Check your price
  • Phospho-PYK2 (Tyr402) Antibody (Oligoclonal, 17HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710087
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (4-5 ug/ml)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
710087 309.00
Check your price
  • CaMKII gamma Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529648
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CAMK, CAMK-II, CAMKG, caMK-II subunit gamma, calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529648 349.00
Check your price