1 - 2 of 2 products displayed
  • AP3B1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529858
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Clathrin assembly protein complex 3 beta-1 large chain; Beta-3A-adaptin; Adaptor-related protein complex 3 subunit beta-1; Adaptor protein complex AP-3 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529858 349.00
Check your price
  • beta-2 Adaptin Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11066
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11066 349.00
Check your price