1 - 2 of 2 products displayed
  • ADORA2B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518422
  • Size 100 µg
Applications
Immunohistochemistry (3µg/ml)
Alternate names:
ADORA2B; adenosine A2b receptor; HGNC:264; ADORA2; netrin-1 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518422 349.00
Check your price
  • ADORA2B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533322
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
ADORA2B, ADORA2, A2b receptor, Adenosine A2b receptor, Adenosine a2b, Adenosine receptor A2b, A2b adenosine receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533322 349.00
Check your price