1 - 7 of 7 products displayed
  • IBA1 Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518039
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
Ionized calcium-binding adapter molecule 1; Microglia response factor; MRF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518039 349.00
Check your price
  • IBA1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523952
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523952 349.00
Check your price
  • IBA1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521274
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521274 349.00
Check your price
  • IBA1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527436
  • Size 100µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527436 349.00
Check your price
  • IBA1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518488
  • Size 100µg
Applications
Immunohistochemistry (2-4 µg/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518488 349.00
Check your price
  • IBA1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518519
  • Size 100µg
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518519 349.00
Check your price
  • IBA1 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA140227
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:50)
Alternate names:
Protein G1; Ionized calcium-binding adapter molecule 1; Microglia response factor; MRF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140227 600.00
Check your price