1 - 30 of 30 products displayed
  • Intestinal Alkaline Phosphatase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529445
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
IAP, Kasahara isozyme, alkaline phosphomonoesterase, glycerophosphatase, intestinal alkaline phosphatase, intestinal-type alkaline phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529445 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514064
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:40-1:80)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514064 309.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512694
  • Size 500 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (1:200), Western Blot (1:250-1:1000)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512694 309.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522210
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
IAP, Kasahara isozyme, alkaline phosphomonoesterase, glycerophosphatase, intestinal alkaline phosphatase, intestinal-type alkaline phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522210 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514544
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514544 349.00
Check your price
  • Intestinal Alkaline Phosphatase Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # HYB2520802
  • Size 200 µl
Applications
ELISA (1:20,000)
Alternate names:
AP, Akp2, Alkaline phosphatase intestinal, Alkaline phosphomonoesterase, ALPI, Glycerophosphatase, IAP, Kasahara isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB2520802 425.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody, Biotin conjugate for IHC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA512691
  • Size 500 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (1:200)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512691 375.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516392
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516392 349.00
Check your price
  • Placental Alkaline Phosphatase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522270
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522270 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119354
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119354 249.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP conjugate for Western Blot, IHC (F), ELISA and IM
  • Host Goat
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # PA126878
  • Size 1 mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (Frozen) (Assay Dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay Dependent)
Alternate names:
IAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126878 375.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP conjugate for Western Blot, IHC (F), ELISA and IM
  • Host Sheep
  • Target Species Bovine
  • Conjugate HRP
  • Catalog # PA126880
  • Size 1 mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (Frozen) (Assay Dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay Dependent)
Alternate names:
IAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126880 309.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody, HRP conjugate for Western Blot, ELISA and IM
  • Host Rabbit
  • Target Species Human
  • Conjugate HRP
  • Catalog # PA126873
  • Size 1 mg
Applications
ELISA (1:1000-1:5000), Immunomicroscopy (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126873 309.00
Check your price
  • Intestinal Alkaline Phosphatase Polyclonal Antibody, Biotin conjugate for Western Blot, ELISA and IM
  • Host Rabbit
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA126872
  • Size 1 mg
Applications
ELISA (1:4000-1:20,000), Immunomicroscopy (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126872 349.00
Check your price
  • Placental Alkaline Phosphatase Polyclonal Antibody for ELISA and IM
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA185170
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunomicroscopy (Assay Dependent)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185170 309.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514672
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514672 349.00
Check your price
  • Placental Alkaline Phosphatase Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129125
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:50), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:500)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129125 309.00
Check your price
  • Intestinal Alkaline Phosphatase Monoclonal Antibody for IHC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MA514742
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
PLAP-1, Regan isozyme, alkaline phosphatase Regan isozyme, alkaline phosphatase, placental type, alkaline phosphomonoesterase, glycerophosphatase, placental alkaline phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514742 349.00
Check your price
  • Intestinal Alkaline Phosphatase Monoclonal Antibody for IHC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MIA1703
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
PLAP-1, Regan isozyme, alkaline phosphatase Regan isozyme, alkaline phosphatase, placental type, alkaline phosphomonoesterase, glycerophosphatase, placental alkaline phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIA1703 600.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515652
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515652 349.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIA1802
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIA1802 600.00
Check your price
  • Placental Alkaline Phosphatase Monoclonal Antibody for IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120245
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (10 µg/ml), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120245 349.00
Check your price
  • Placental Alkaline Phosphatase Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530311
  • Size 100 µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530311 349.00
Check your price
  • ALPL Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547419
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547419 375.00
Check your price
  • ALPL Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521332
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521332 349.00
Check your price
  • PLAP Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA532554
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:40)
Alternate names:
Placental alkaline phosphatase 1; Alkaline phosphatase Regan isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532554 199.00
Check your price
  • Alkaline Phosphatase Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # ME6200
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Green Renilla luciferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
ME6200 185.00
Check your price
  • Alkaline Phosphatase Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # 459260
  • Size 2.5 mL
Applications
Immunocytochemistry (1:2-1:3.), Immunofluorescence (Assay Dependent), Western Blot (1:50 to 1:100)
Alternate names:
AP
Catalog # Size Price (AUD) Your Price (AUD) Qty
459260 Not Orderable Online
Check your price
  • Alkaline Phosphatase Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA180405
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin, Frozen) (1/30-1/50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180405 375.00
Check your price
  • ALPL Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA135004
  • Size 100 µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135004 375.00
Check your price