1 - 30 of 30 products displayed
  • CK2 alpha-1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528686
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CK2A1, CKII, CK II alpha, CK2 catalytic subunit alpha, casein kinase II alpha 1 subunit, casein kinase II subunit alpha, protein kinase CK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528686 349.00
Check your price
  • ATP1A1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3929
  • Size 100µg
Applications
Flow Cytometry (1/20-1/100), Immunocytochemistry (1/50-1/200), Immunofluorescence (1/50-1/200), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1/100-1/2000), Western Blot (1:200-1:2000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3929 375.00
Check your price
  • Phospho-Sodium / Potassium ATPase alpha-1 pSer23 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535781
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535781 Not Orderable Online
Check your price
  • Calcium Channel 1A Polyclonal Antibody for Western Blot, IHC (F) and IHC (PFA)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519598
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:100), Western Blot (Assay-Dependent)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav2.1; Rat brain class A; Calcium channel, L type, alpha-1 polypeptide, isoform 4; Brain calcium channel I; Calcium channel, L type, alpha-1 polypeptide isoform 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519598 349.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA122860
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor; Alpha-1-proteinase inhibitor; Alpha-1-antitrypsin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA122860 309.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # PA126909
  • Size 1mg
Applications
ELISA (1:50,000), Western Blot (1:2000-1:10,000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126909 375.00
Check your price
  • alpha-1 Antitrypsin Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121205
  • Size 250µg
Applications
ELISA (1:1000-1:5000), Western Blot (1:1000-1:5000)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121205 375.00
Check your price
  • Dihydropyridine Receptor alpha-1 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3920
  • Size 200µL
Applications
Flow Cytometry (1ug/test), Immunohistochemistry (Frozen) (1:200), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav1.1; Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3920 349.00
Check your price
  • Dihydropyridine Receptor alpha-1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521464
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Voltage-gated calcium channel subunit alpha Cav1.1; Calcium channel, L type, alpha-1 polypeptide, isoform 3, skeletal muscle
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521464 349.00
Check your price
  • Collagen II Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA126206
  • Size 100µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126206 349.00
Check your price
  • COL6A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529068
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
OPLL, alpha 1 (VI) chain (61 AA), collagen VI, alpha-1 polypeptide, collagen alpha-1(VI) chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529068 349.00
Check your price
  • Phospho-ATP1A1 (Ser16) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536732
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536732 349.00
Check your price
  • Collagen II Polyclonal Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Biotin
  • Catalog # PA128531
  • Size 50µg
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:5000-1:10,000)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128531 349.00
Check your price
  • alpha/beta Tubulin Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA186602
  • Size 500µL
Applications
ELISA (1/50-1/500), Western Blot (1/250-1/1000)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin beta-5 chain; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186602 349.00
Check your price
  • Phospho-ATP1A1 (Ser16) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538261
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538261 309.00
Check your price
  • alpha/beta Tubulin Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA119399
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Western Blot (5 ug/ml)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin beta-5 chain; Tubulin alpha-1 chain; Alpha-tubulin isotype M-alpha-1; Alpha-tubulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119399 225.00
Check your price
  • alpha/beta Tubulin Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA119400
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin beta-5 chain; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119400 309.00
Check your price
  • Phospho-RSK1 (Thr348) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537730
  • Size 100µl
Applications
Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537730 309.00
Check your price
  • Collagen IV Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA128534
  • Size 100µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:5000-1:10,000), Western Blot (1:5000-1:10,000)
Alternate names:
arresten
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128534 375.00
Check your price
  • Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537829
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
pp90RSK3; Ribosomal S6 kinase 3; MAP kinase-activated protein kinase 1c; 90 kDa ribosomal protein S6 kinase 2; pp90RSK2; Ribosomal S6 kinase 2; MAP kinase-activated protein kinase 1b; Insulin-stimulated protein kinase 1; 90 kDa ribosomal protein S6 kinase 3; Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1; pp90RSK4; Ribosomal S6 kinase 4; 90 kDa ribosomal protein S6 kinase 6; SAPK-beta; Protein-tyrosine kinase Mpk-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537829 309.00
Check your price
  • Phospho-RSK1 (Ser352) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537729
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537729 309.00
Check your price
  • Phospho-RSK1 (Thr359, Ser363) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538309
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538309 309.00
Check your price
  • Phospho-RSK1 (Thr573) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537728
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537728 309.00
Check your price
  • COL8A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537109
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Endothelial collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537109 309.00
Check your price
  • CK2 alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528602
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CK2A1, CKII, CK II alpha, CK2 catalytic subunit alpha, casein kinase II alpha 1 subunit, casein kinase II subunit alpha, protein kinase CK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528602 349.00
Check your price
  • CK2 alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529185
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
CK2A2, CSNK2A1, CK II alpha', casein kinase II subunit alpha'
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529185 349.00
Check your price
  • ATP1A1 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3928
  • Size 400µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:20), Immunohistochemistry (1:100), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:5000)
Alternate names:
Sodium pump subunit alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3928 375.00
Check your price
  • Phospho-CK2 alpha (Thr360, Ser362) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537540
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
CK2A1, CKII, CK II alpha, CK2 catalytic subunit alpha, casein kinase II alpha 1 subunit, casein kinase II subunit alpha, protein kinase CK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537540 309.00
Check your price
  • Phospho-CK2 alpha (Tyr255) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538831
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
CK2A1, CKII, CK II alpha, CK2 catalytic subunit alpha, casein kinase II alpha 1 subunit, casein kinase II subunit alpha, protein kinase CK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538831 309.00
Check your price
  • alpha-1 Antitrypsin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB1910102
  • Size 200µl
Applications
ELISA (1:15,000), Western Blot (Assay dependent)
Alternate names:
Serpin A1; Alpha-1-antiproteinase; Alpha-1 protease inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1910102 425.00
Check your price