1 - 30 of 30 products displayed
  • Donkey anti-Chicken IgY Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Donkey
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # SA172004
  • Size 500µg
Applications
ELISA (1:15000), Western Blot (1:7500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172004 149.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Texas Red® conjugate for IHC (F), IHC (P), Flow and IM
  • Host Goat
  • Target Species Chicken
  • Conjugate Texas Red®
  • Catalog # PA128656
  • Size 1mg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128656 197.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Texas Red® conjugate for IHC (F) and IHC (P)
  • Host Goat
  • Target Species Chicken
  • Conjugate Texas Red®
  • Catalog # PA128796
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128796 151.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # PA128797
  • Size 100µg
Applications
ELISA (1:8000-1:36000), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128797 168.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # PA128657
  • Size 1mg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128657 209.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, TRITC conjugate for IHC (F), IHC (P), Flow and IM
  • Host Goat
  • Target Species Chicken
  • Conjugate TRITC
  • Catalog # PA128795
  • Size 100µg
Applications
Flow Cytometry (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay-dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128795 170.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, TRITC conjugate for Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate TRITC
  • Catalog # PA128655
  • Size 1mg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128655 194.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, FITC conjugate for Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # PA128794
  • Size 100µg
Applications
Flow Cytometry (1:2000-1:10,000), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (1:500-1:2500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128794 147.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # PA128798
  • Size 100µg
Applications
ELISA (1:10,000-1:50,000), Immunohistochemistry (Paraffin, Frozen) (1:500-1:2500), Immunomicroscopy (Assay Dependent), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128798 168.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA, DB and IM
  • Host Goat
  • Target Species Chicken
  • Conjugate AP
  • Catalog # PA128799
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (1:1000-1:4000), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunomicroscopy (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128799 147.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC (F), IHC (P), ELISA and IM
  • Host Goat
  • Target Species Chicken
  • Conjugate AP
  • Catalog # PA130369
  • Size 1mg
Applications
ELISA (1:4000-1:16,000), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:200-1:1000), Immunomicroscopy (Assay-dependent), Western Blot (1:1000-1:4000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130369 383.00
Check your price
  • Goat anti-Chicken IgY Fab Secondary Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # SA172005
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172005 131.00
Check your price
  • Goat anti-Chicken IgY Fc Secondary Antibody, HRP conjugate for Western Blot and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # SA172012
  • Size 1mL
Applications
ELISA (0.2-0.05 ug/ml), Western Blot (0.2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172012 184.00
Check your price
  • Goat anti-Chicken IgY Fc Secondary Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # SA172009
  • Size 500µg
Applications
ELISA (5 ug/ml), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172009 163.00
Check your price
  • Donkey anti-Chicken IgY Secondary Antibody, FITC conjugate for Western Blot and ELISA
  • Host Donkey
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # SA172000
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172000 189.00
Check your price
  • Donkey anti-Chicken IgY Secondary Antibody for Western Blot and ELISA
  • Host Donkey
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # SA172002
  • Size 500µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172002 131.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Qdot® 655 conjugate for Western Blot, ICC, IHC and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate Qdot® 655
  • Catalog # Q14421MP
  • Size 200µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50), Immunohistochemistry (1:50), Western Blot (1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
Q14421MP 311.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate
  • Catalog # A16134
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16134 161.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Rabbit
  • Target Species Chicken
  • Conjugate TRITC
  • Catalog # A16135
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16135 96.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Rabbit
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # A16131
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16131 136.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, Biotin conjugate for IF, Flow and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # A16058
  • Size 2mg
Applications
ELISA (1:500-1:20,000), Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16058 103.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, TRITC conjugate for IF
  • Host Goat
  • Target Species Chicken
  • Conjugate TRITC
  • Catalog # A16059
  • Size 1mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16059 94.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # A16130
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16130 149.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, FITC conjugate for IF and Flow
  • Host Goat
  • Target Species Chicken
  • Conjugate FITC
  • Catalog # A16055
  • Size 2mg
Applications
Flow Cytometry (1:500-1:20,000), Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16055 102.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate
  • Catalog # A16133
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16133 Not Orderable Online
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate HRP
  • Catalog # A16054
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16054 109.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody, AP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate AP
  • Catalog # A16057
  • Size 1mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16057 179.00
Check your price
  • Rabbit anti-Chicken IgY (H+L) Secondary Antibody for ELISA
  • Host Rabbit
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # A16132
  • Size 2mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16132 157.00
Check your price
  • Goat anti-Chicken IgY (H+L) Secondary Antibody for ELISA
  • Host Goat
  • Target Species Chicken
  • Conjugate Unconjugated
  • Catalog # A16056
  • Size 2mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16056 95.25
Check your price
  • Donkey anti-Chicken IgY Secondary Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Donkey
  • Target Species Chicken
  • Conjugate Biotin
  • Catalog # SA172003
  • Size 500µg
Applications
ELISA (1:10000-1:20000), Western Blot (1:1000-1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA172003 162.00
Check your price