1 - 20 of 20 products displayed
  • eGFP Tag Antibody (Monoclonal, F56-6A1.2.3)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1952
  • Size 100 µg
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100), Western Blot (1:1,000)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1952 169.00
Check your price
  • eGFP Tag Antibody
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # CAB4211
  • Size 100 ug
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Western Blot (1 ug/ml)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
CAB4211 159.00
Check your price
  • GFP Tag Antibody (Monoclonal, GF28R)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated See Additional Formats Biotin DyLight 650 DyLight 680 DyLight 800 4X PEG HRP
  • Catalog # MA515256
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:500-1:2000), Immunoprecipitation (Assay Dependent), Western Blot (1:1000-1:3000)
Alternate names:
Green Fluorescent Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515256 169.00
Check your price
  • GFP Tag Antibody, Alexa Fluor® 594
  • Host Rabbit
  • Target Species Tag
  • Conjugate Alexa Fluor® 594
  • Catalog # A21312
  • Size 100 µL
Applications
Immunocytochemistry (1-10 ug/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21312 349.00
Check your price
  • GFP Tag Antibody (Monoclonal, 3E6)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # A11120
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11120 299.00
Check your price
  • GFP Tag Antibody
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # A11122
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11122 349.00
Check your price
  • GFP Tag Antibody (Monoclonal, C163)
  • Host Mouse
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # 332600
  • Size 100 µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
332600 249.00
Check your price
  • TurboGFP Antibody
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # PA522688
  • Size 100 µg
Applications
ELISA (1:30,000-1:50,000), Immunocytochemistry (1:5,000-1:10,000), Immunofluorescence (1:5000-1:10,000), Western Blot (1:5000-1:10000)
Alternate names:
GFP2, Green Fluoresecent Protein, GFP, Turbo GFP, CopGFP, ppluGFP2,
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522688 349.00
Check your price
  • Goat anti-Mouse IgG (H+L), DyLight® 550
  • Host Goat
  • Target Species Mouse
  • Conjugate DyLight® 550
  • Catalog # 84540
  • Size 1 mL
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
84540 155.00
Check your price
  • Goat anti-Rabbit IgG (H+L), DyLight® 550
  • Host Goat
  • Target Species Rabbit
  • Conjugate DyLight® 550
  • Catalog # 84541
  • Size 1 mL
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
84541 155.00
Check your price
  • SMURF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511946
  • Size 400 µL
Applications
Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
SMAD-specific E3 ubiquitin-protein ligase 2; SMAD ubiquitination regulatory factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511946 325.00
Check your price
  • ABRA1 Antibody (Monoclonal, ABRA1-01)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110045
  • Size 100 ug
Applications
Flow Cytometry (1-2 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein FAM175A; Coiled-coil domain-containing protein 98
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110045 249.00
Check your price
  • MRCK alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA110038
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (1:200)
Alternate names:
Myotonic dystrophy kinase-related CDC42-binding kinase alpha; DMPK-like alpha; CDC42-binding protein kinase alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110038 275.00
Check your price
  • ZNF322 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Goat, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA541636
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Zinc finger protein 322A; Zinc finger protein 489; Zinc finger protein 388
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541636 309.00
Check your price
  • CEACAM6 Antibody (Monoclonal, 439424)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524164
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524164 299.00
Check your price
  • HMBOX1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541634
  • Size 100 µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Homeobox containing 1, PBHNF, HNF1LA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541634 309.00
Check your price
  • PEA15 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544950
  • Size 100 µl
Applications
Western Blot (1:1000)
Alternate names:
15 kDa phosphoprotein enriched in astrocytes; Phosphoprotein enriched in diabetes
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544950 309.00
Check your price
  • PEA15 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543145
  • Size 100 µl
Applications
Western Blot (1:1000)
Alternate names:
15 kDa phosphoprotein enriched in astrocytes; Phosphoprotein enriched in diabetes
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543145 309.00
Check your price
  • LINGO2 Antibody (Monoclonal, 382007)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524121
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Leucine-rich repeat neuronal protein 6C; Leucine-rich repeat neuronal protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524121 299.00
Check your price
  • SLAMF8 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547220
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
BCM-like membrane protein; B-lymphocyte activator macrophage expressed
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547220 325.00
Check your price