1 - 30 of 30 products displayed
  • CD34 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517827
  • Size 100ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517827 129.00
Check your price
  • CD34 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517828
  • Size 300ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517828 309.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517829
  • Size 100ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517829 149.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517830
  • Size 300ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517830 309.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517831
  • Size 50ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517831 119.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517832
  • Size 300ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517832 375.00
Check your price
  • CD34 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517825
  • Size 250ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517825 225.00
Check your price
  • CD34 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA517826
  • Size 500ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517826 325.00
Check your price
  • CD34 Polyclonal Antibody for Western Blot and Flow
  • Host Sheep
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547849
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547849 349.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523596
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523596 279.00
Check your price
  • CD34 Monoclonal Antibody, PE-Cyanine7 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine7
  • Catalog # A16238
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16238 138.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A16188
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16188 108.00
Check your price
  • CD34 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # A16226
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16226 102.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A16203
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16203 112.00
Check your price
  • CD34 Monoclonal Antibody for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate
  • Catalog # A14931
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14931 Not Orderable Online
Check your price
  • CD34 Monoclonal Antibody for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate
  • Catalog # A14933
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14933 Not Orderable Online
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119645
  • Size 100tests
Applications
Flow Cytometry (20ul x 100ul whole blood or 10^6 cells)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119645 309.00
Check your price
  • CD34 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119229
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119229 199.00
Check your price
  • CD34 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA516927
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516927 349.00
Check your price
  • CD34 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # A14948
  • Size 25tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14948 Not Orderable Online
Check your price
  • CD34 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547582
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547582 349.00
Check your price
  • CD68 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523572
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Gp110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523572 249.00
Check your price
  • CD68 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523616
  • Size 100Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Gp110
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523616 299.00
Check your price
  • STRO-1 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 398401
  • Size 100µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
398401 305.00
Check your price
  • CD31 Monoclonal Antibody, Alexa Fluor® 647 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 647
  • Catalog # A14716
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
A14716 109.00
Check your price
  • CD31 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A16201
  • Size 50µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
A16201 108.00
Check your price
  • PODXL2 Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547502
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Endoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547502 349.00
Check your price
  • CD31 Monoclonal Antibody, Pacific Blue™ conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Pacific Blue™
  • Catalog # RM5228
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM5228 293.00
Check your price
  • CD31 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 370700
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
370700 304.00
Check your price
  • SIGLEC7 Polyclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547079
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Neutralization (0.3-0.9 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
p75; QA79 membrane protein; D-siglec; CDw328; Adhesion inhibitory receptor molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547079 349.00
Check your price