1 - 30 of 30 products displayed
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 800
  • Catalog # SA510044
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510044 158.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 680 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 680
  • Catalog # SA510042
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510042 158.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 488
  • Catalog # SA510038
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510038 158.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 594 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 594
  • Catalog # SA510040
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510040 158.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 550
  • Catalog # SA510039
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510039 157.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 800 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate DyLight® 800
  • Catalog # SA510036
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510036 158.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 550 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate DyLight® 550
  • Catalog # SA510033
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510033 156.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 650
  • Catalog # SA510041
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510041 158.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 755 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 755
  • Catalog # SA510043
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510043 157.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 350 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Donkey
  • Target Species Rabbit
  • Conjugate DyLight® 350
  • Catalog # SA510037
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510037 156.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 755 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate DyLight® 755
  • Catalog # SA510035
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510035 156.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight® 650 conjugate for Western Blot, IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate DyLight® 650
  • Catalog # SA510034
  • Size 500 ug
Applications
Flow Cytometry (1:25-1:100), Immunocytochemistry (1:50-1:2,000), Immunofluorescence (1:50-1:2,000), Immunohistochemistry (1:50-1:2,000), Immunoprecipitation (Assay Dependent), Western Blot (1:5,000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA510034 156.00
Check your price
  • Donkey anti-Rabbit IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Donkey
  • Target Species Rabbit
  • Conjugate WesternDot® 625
  • Catalog # W10806
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10806 183.00
Check your price
  • Donkey anti-Rabbit IgG Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Donkey
  • Target Species Rabbit
  • Conjugate WesternDot® 655
  • Catalog # W10811
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10811 183.00
Check your price
  • Donkey anti-Rabbit IgG Secondary Antibody, WesternDot® 585 conjugate for Western Blot
  • Host Donkey
  • Target Species Rabbit
  • Conjugate WesternDot® 585
  • Catalog # W10801
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10801 183.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 625
  • Catalog # W10809
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10809 183.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 585 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 585
  • Catalog # W10804
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10804 184.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 655 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 655
  • Catalog # W10814
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10814 184.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 800
  • Catalog # W10816
  • Size 25 blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10816 181.00
Check your price
  • Donkey anti-Rabbit IgG Secondary Antibody, WesternDot® 800 conjugate for Western Blot
  • Host Donkey
  • Target Species Rabbit
  • Conjugate WesternDot® 800
  • Catalog # W10824
  • Size 25 blots
Applications
Western Blot (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10824 181.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, APC conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate APC
  • Catalog # A10931
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10931 309.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 680
  • Catalog # A21109
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL), Western Blot (0.04-0.2 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21109 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 680 conjugate for Western Blot, IF and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 680
  • Catalog # A21076
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21076 169.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 633
  • Catalog # A21070
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21070 169.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 633 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 633
  • Catalog # A21071
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21071 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 568 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 568
  • Catalog # A11036
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11036 209.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 555
  • Catalog # A21428
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21428 169.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor® 555 conjugate for IF, ICC and Flow
  • Host Goat
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 555
  • Catalog # A21429
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21429 209.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # 31636
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31636 173.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, APC conjugate for IF, ICC, IHC, Flow and IP
  • Host Goat
  • Target Species Rabbit
  • Conjugate APC
  • Catalog # 31984
  • Size 500 µL
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31984 222.00
Check your price