1 - 30 of 30 products displayed
  • Actin Antibody (Monoclonal, mAbGEa)
  • Host Mouse
  • Target Species Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA1744
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Cytoplasmic beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1744 375.00
Check your price
  • Actin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511570
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Beta-actin; Alpha-cardiac actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511570 349.00
Check your price
  • gamma Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513467
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Gamma-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513467 349.00
Check your price
  • ACTR3B Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544774
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Actin-like protein 3B; ARP3-beta; Actin-related protein ARP4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544774 309.00
Check your price
  • PHACTR1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544332
  • Size 100 µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Phosphatase and actin regulator 1, KIAA1733, MGC126575, MGC126577, RPEL, RPEL1, dJ257A7.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544332 309.00
Check your price
  • Phospho-p38 MAPK alpha (Thr180, Tyr182) Antibody (Monoclonal, C.7.8)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA515218
  • Size 100 µL
Applications
Flow Cytometry (1:800), Immunocytochemistry (1:400), Immunofluorescence (1:100-1:500), Immunoprecipitation (1:50), Western Blot (1:2000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; Mitogen-activated protein kinase p38a; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515218 309.00
Check your price
  • VPS52 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524408
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
SAC2 suppressor of actin mutations 2-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524408 349.00
Check your price
  • ACTR1A Antibody (Monoclonal, 3E5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520935
  • Size 100 µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Centrosome-associated actin homolog; Actin-RPV; ARP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520935 309.00
Check your price
  • Phospho-PAK1 (Thr423) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512844
  • Size 400 µL
Applications
Dot blot (1:500), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50)
Alternate names:
p65-PAK; p21-activated kinase 1; Alpha-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512844 349.00
Check your price
  • ACTR1B Antibody (Monoclonal, 4E10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520934
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml)
Alternate names:
Actin-related protein 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520934 309.00
Check your price
  • ASF1B Antibody (Monoclonal, K.860.0)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514836
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
CCG1-interacting factor A-II; Anti-silencing function protein 1 homolog B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514836 275.00
Check your price
  • VPS52 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544815
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
SAC2 suppressor of actin mutations 2-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544815 309.00
Check your price
  • PAK1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515115
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
p65-PAK; p21-activated kinase 1; Alpha-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515115 349.00
Check your price
  • Phospho-PAK1/2/3 (Thr402) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA14636
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
p68-PAK; p21-activated kinase 1; Protein kinase MUK2; Alpha-PAK; p65-PAK; p21-activated kinase 3; Beta-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14636 375.00
Check your price
  • SMARCD2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA541441
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-16 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
BRG1-associated factor 60B; 60 kDa BRG-1/Brm-associated factor subunit B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541441 309.00
Check your price
  • BAF60C Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 720131
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
BRG1-associated factor 60C; 60 kDa BRG-1/Brm-associated factor subunit C
Catalog # Size Price (AUD) Your Price (AUD) Qty
720131 309.00
Check your price
  • ACTR6 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA545981
  • Size 100 µl
Applications
Western Blot (1 µg/ml)
Alternate names:
hARPX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545981 309.00
Check your price
  • Phospho-PAK1/2/3 (Ser141) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44940G
  • Size 10 blots
Applications
Western Blot (1-2 µg/mL)
Alternate names:
p21-activated kinase 3; Oligophrenin-3; Beta-PAK; p58; p21-activated kinase 2; S6/H4 kinase; PAK65; Gamma-PAK; p65-PAK; p21-activated kinase 1; Alpha-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
44940G 304.00
Check your price
  • PSMC2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1969
  • Size 100 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:1,000)
Alternate names:
Protein MSS1; Proteasome 26S subunit ATPase 2; 26S proteasome AAA-ATPase subunit RPT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1969 349.00
Check your price
  • ECM29 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3035
  • Size 100 µl
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:200)
Alternate names:
FLJ22036, KIAA1962, Proteasome associated protein ECM29 homolog, RP11-386D8.2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3035 349.00
Check your price
  • SMARCB1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA540834
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
SNF5 homolog; Integrase interactor 1 protein; BRG1-associated factor 47
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540834 309.00
Check your price
  • PAK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 719300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
p68-PAK; p21-activated kinase 1; Protein kinase MUK2; Alpha-PAK; p65-PAK
Catalog # Size Price (AUD) Your Price (AUD) Qty
719300 309.00
Check your price
  • MST3 Antibody (Monoclonal, ZM003)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 396200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
STE20-like kinase MST3; Mammalian STE20-like protein kinase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
396200 309.00
Check your price
  • ATG12 Antibody (Oligoclonal, 20 HCLC)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710716
  • Size 100 µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Immunocytochemistry (2 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (2.5-4 µg/ml)
Alternate names:
Autophagy-related protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
710716 309.00
Check your price
  • eIF1 Antibody (Monoclonal, 2B9)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1077
  • Size 100 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
Sui1iso1; Protein translation factor SUI1 homolog; A121
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1077 349.00
Check your price
  • alpha Tubulin Antibody (Monoclonal, YOL1/34)
  • Host Rat
  • Target Species Dog, Human, Mouse, Non-human primate, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA180189
  • Size 500 µg
Applications
ELISA (10ug/ml as detecting anti), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tuba1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180189 349.00
Check your price
  • alpha Tubulin Antibody (Monoclonal, YL1/2)
  • Host Rat
  • Target Species Dog, Fruit fly, Human, Mouse, Plant, Pig, Rabbit, Rat, Xenopus, Yeast
  • Conjugate Unconjugated
  • Catalog # MA180017
  • Size 500 µg
Applications
ELISA (1/100-1/1000), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tubulin alpha-3 chain; Tubulin B-alpha-1; Alpha-tubulin 3; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180017 399.00
Check your price
  • beta Tubulin Antibody
  • Host Rabbit
  • Target Species Amphibian, Bovine, Chicken, Fungi, Gerbil, Guinea pig, Human, Mouse, Pig, Rat, Sea urchin, Yeast
  • Conjugate Unconjugated
  • Catalog # PA516863
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tubulin beta-5 chain; Tubulin beta 4'
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516863 309.00
Check your price
  • Annexin A1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 713400
  • Size 100 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:500-1:2000)
Alternate names:
p35; Phospholipase A2 inhibitory protein; Lipocortin I; Chromobindin-9; Calpactin-2; Calpactin II; Annexin-1; Annexin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
713400 307.00
Check your price
  • Lactoferrin Antibody (Monoclonal, 1B8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0109
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (1-2 µl)
Alternate names:
Talalactoferrin; Growth-inhibiting protein 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0109 309.00
Check your price