1 - 30 of 30 products displayed
  • TBP Monoclonal Antibody for Western Blot, IF and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1189
  • Size 100µl
Applications
Flow Cytometry (1-20µg/ml), Immunofluorescence (1-20µg/ml), Western Blot (1:1000)
Alternate names:
Transcription initiation factor TFIID TBP subunit; TATA-box factor; TATA-binding factor; TATA sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1189 309.00
Check your price
  • E2F1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123208
  • Size 100µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay-dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
pRB-binding protein E2F-1; Retinoblastoma-binding protein 3; Retinoblastoma-associated protein 1; PBR3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123208 309.00
Check your price
  • E2F1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123202
  • Size 100µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1-5 µg/ml)
Alternate names:
pRB-binding protein E2F-1; Retinoblastoma-binding protein 3; Retinoblastoma-associated protein 1; PBR3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123202 309.00
Check your price
  • SSBP1 Polyclonal Antibody for Western Blot, IF, ICC
  • Host Chicken
  • Target Species Mouse, Human, Rat
  • Conjugate
  • Catalog # PA127871
  • Size 50µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (1:2000)
Alternate names:
Single strand DNA-binding protein P16; PWP1-interacting protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127871 Not Orderable Online
Check your price
  • p300 Polyclonal Antibody for Western Blot and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1848
  • Size 100µg
Applications
ChIP assay (1-3 ul), Western Blot (1-2 µg/ml)
Alternate names:
E1A-associated protein p300
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1848 349.00
Check your price
  • CBP Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1847
  • Size 100µg
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:1000-1:5000), Western Blot (1-2 µg/ml)
Alternate names:
CREB Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1847 375.00
Check your price
  • CHP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA529876
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p24; Sid 470; EF-hand calcium-binding domain-containing protein p22; Calcium-binding protein p22; Calcium-binding protein CHP; Calcineurin B-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529876 349.00
Check your price
  • NR5A2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534018
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (10-19 µg/ml)
Alternate names:
Liver receptor homolog 1; Hepatocytic transcription factor; CYP7A promoter-binding factor; B1-binding factor; Alpha-1-fetoprotein transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534018 349.00
Check your price
  • NR5A2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534017
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (7 µg/ml)
Alternate names:
Liver receptor homolog 1; Hepatocytic transcription factor; CYP7A promoter-binding factor; B1-binding factor; Alpha-1-fetoprotein transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534017 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534164
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (6 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534164 349.00
Check your price
  • NR5A1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124565
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (Assay dependent)
Alternate names:
Steroid hormone receptor Ad4BP; Nuclear receptor subfamily 5 group A member 1; Fushi tarazu factor homolog 1; Adrenal 4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124565 399.00
Check your price
  • NR5A1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534015
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (7 µg/ml)
Alternate names:
Steroid hormone receptor Ad4BP; Nuclear receptor subfamily 5 group A member 1; Fushi tarazu factor homolog 1; Adrenal 4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534015 349.00
Check your price
  • NR5A1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130599
  • Size 50µg
Applications
Western Blot (1-5 µg/ml)
Alternate names:
Steroid hormone receptor Ad4BP; Nuclear receptor subfamily 5 group A member 1; Fushi tarazu factor homolog 1; Adrenal 4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130599 425.00
Check your price
  • PAG1 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA119287
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay-Dependent)
Alternate names:
Transmembrane phosphoprotein Cbp; Csk-binding protein; Transmembrane adapter protein PAG; Pregnancy-specific protein B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119287 225.00
Check your price
  • TBP Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA514739
  • Size 100µg
Applications
ChIP assay (5-10 µg for 25 µg of chromatin), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
Transcription initiation factor TFIID TBP subunit; TATA-box factor; TATA-binding factor; TATA sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514739 349.00
Check your price
  • TBP Polyclonal Antibody for Western Blot, IF, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519776
  • Size 100µg
Applications
ChIP assay (5 µg for 25 µg of chromatin), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Transcription initiation factor TFIID TBP subunit; TATA-box factor; TATA-binding factor; TATA sequence-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519776 349.00
Check your price
  • DBP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124453
  • Size 50µg
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (0.5-5 µg/ml)
Alternate names:
Tax-responsive enhancer element-binding protein 302; Albumin D-element-binding protein; Albumin D box-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124453 425.00
Check your price
  • GAL4 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # 338600
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Western Blot (Assay Dependent)
Alternate names:
GAL4, L36LBP, L-36 lactose-binding protein, antigen NY-CO-27, gal-4, galectin 4, galectin-4, lactose-binding lectin 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
338600 223.00
Check your price
  • CREB Monoclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701120
  • Size 100µg
Applications
ChIP assay (1 ul), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Western Blot (1:500-1:5000)
Alternate names:
Active transcription factor CREB, cAMP responsive element binding protein 1, CREB1, MGC9284, Transactivator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701120 309.00
Check your price
  • GAL4 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA3031
  • Size 50µl
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:2000)
Alternate names:
Lactose-binding lectin 4; L-36 lactose-binding protein; Antigen NY-CO-27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3031 309.00
Check your price
  • GATA2 Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # 710242
  • Size 100µg
Applications
Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (2-5 ug/ml)
Alternate names:
GATA-binding protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710242 309.00
Check your price
  • FABP4 Oligoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710189
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710189 309.00
Check your price
  • FABP4 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701158
  • Size 100µg
Applications
Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (0.1-1 ug/ml)
Alternate names:
Fatty acid-binding protein 4; Adipocyte-type fatty acid-binding protein; Adipocyte lipid-binding protein; Protein 422; P2 adipocyte protein; P15; Myelin P2 protein homolog; 3T3-L1 lipid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701158 304.00
Check your price
  • RORB Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534160
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
RORB, BA133M9.1, Nuclear receptor ROR-beta, NR1F2, ROR Beta, ROR-BETA, Rorbeta, RZR-BETA, Nuclear receptor RZR-beta, RAR-related orphan receptor B, RZRB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534160 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534167
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534167 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534166
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (22 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534166 349.00
Check your price
  • RORC Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534165
  • Size 50ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (15-20 µg/ml)
Alternate names:
Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534165 349.00
Check your price
  • GC Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529082
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Vitamin D-binding protein-macrophage activating factor; Group-specific component; Gc-globulin; Gc protein-derived macrophage activating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529082 349.00
Check your price
  • GC Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529067
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Vitamin D-binding protein-macrophage activating factor; Group-specific component; Gc-globulin; Gc protein-derived macrophage activating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529067 349.00
Check your price
  • GC Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519802
  • Size 100µg
Applications
Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Vitamin D-binding protein-macrophage activating factor; Group-specific component; Gc-globulin; Gc protein-derived macrophage activating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519802 349.00
Check your price