1 - 3 of 3 products displayed
  • APOE Antibody (Monoclonal, 4E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518068
  • Size 100 µL
Applications
Dot blot (Assay-dependent), ELISA (Assay-dependent), Flow Cytometry (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:64,000)
Alternate names:
AD2; LDLCQ5; LPG; apo-E; apolipoprotein E4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518068 425.00
Check your price
  • APOE Antibody (Monoclonal, 4E4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516146
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunoprecipitation (1:10-1:500), Western Blot (1:1000-1:2000)
Alternate names:
AD2, LDLCQ5, LPG, apo-E, apolipoprotein E3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516146 349.00
Check your price
  • APOE Antibody (Monoclonal, E6D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191063
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
AD2, LDLCQ5, LPG, MGC1571
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191063 550.00
Check your price