1 - 30 of 30 products displayed
  • LXR beta Antibody (Monoclonal, K8917)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 418400
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
418400 433.00
Check your price
  • PIK3R1 Antibody (Monoclonal, U13)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA121472
  • Size 1 mL
Applications
Immunoprecipitation (Neat), Western Blot (1:200)
Alternate names:
Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121472 349.00
Check your price
  • HNF4A Antibody (Monoclonal, H1415)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417700
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
417700 428.00
Check your price
  • HNF4A Antibody (Monoclonal, H6939)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Transcription factor HNF-4; Transcription factor 14; Nuclear receptor subfamily 2 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
417800 433.00
Check your price
  • TRF2 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141023
  • Size 100 µg
Applications
ChIP assay (1:10-1:500), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (15 µg/ml), Immunoprecipitation (2 µg/10^6 cells), Western Blot (2 µg/ml)
Alternate names:
Telomeric DNA-binding protein; TTAGGG repeat-binding factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141023 349.00
Check your price
  • Hepatitis B Virus X Antibody (Monoclonal, X36C)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA1081
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
HBV x Core Protein, HBVgp3, HepB X
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1081 375.00
Check your price
  • NuMA Antibody
  • Host Rabbit
  • Target Species Human, Mammal
  • Conjugate Unconjugated
  • Catalog # PA132451
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (1:500), Immunoprecipitation (Assay dependent), Western Blot (1:1000)
Alternate names:
SP-H antigen; Nuclear matrix protein-22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132451 399.00
Check your price
  • PPAR gamma Antibody (Monoclonal, A3409A)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 419300
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group C member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
419300 426.00
Check your price
  • NR4A1 Antibody (Monoclonal, H1648)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 418600
  • Size 100 µg
Applications
ELISA (0.3µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Orphan nuclear receptor HMR; Nuclear protein N10; Nuclear hormone receptor NUR/77; Testicular receptor 3; ST-59; Orphan nuclear receptor TR3; Early response protein NAK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
418600 433.00
Check your price
  • CDK5 Antibody (Monoclonal, 8A1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0116
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunoprecipitation (2 µl), Western Blot (1:1000-1:2000)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0116 309.00
Check your price
  • CDK5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # LFPA0132
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
Tau protein kinase II catalytic subunit; Serine/threonine-protein kinase PSSALRE; Cell division protein kinase 5; CR6 protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFPA0132 309.00
Check your price
  • PTPN12 Antibody (Monoclonal, AG25)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA112377
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein-tyrosine phosphatase P19; MPTP-PEST; Protein-tyrosine phosphatase G1; PTP-PEST
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112377 309.00
Check your price
  • Estrogen Receptor alpha Antibody (Monoclonal, H4624)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 417000
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
417000 433.00
Check your price
  • Androgen Receptor Antibody (Monoclonal, H7507)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 415900
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group C member 4; Dihydrotestosterone receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
415900 433.00
Check your price
  • RXRA Antibody (Monoclonal, K8505)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 433900
  • Size 100 µg
Applications
ChIP assay (Assay Dependent), ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor alpha; Nuclear receptor subfamily 2 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
433900 426.00
Check your price
  • LXR alpha Antibody (Monoclonal, K8607)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 418200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
418200 433.00
Check your price
  • TLX Antibody (Monoclonal, H6510)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 434500
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein tailless homolog; Nuclear receptor TLX
Catalog # Size Price (AUD) Your Price (AUD) Qty
434500 425.00
Check your price
  • EHMT2 Antibody (Monoclonal, A8620A)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 435200
  • Size 100 µg
Applications
ELISA (0.1 µg/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (5 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Protein G9a; Lysine N-methyltransferase 1C; Histone H3-K9 methyltransferase 3; HLA-B-associated transcript 8; Euchromatic histone-lysine N-methyltransferase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
435200 427.00
Check your price
  • FXR Antibody (Monoclonal, A9033A)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 417200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor-interacting protein 14; Nuclear receptor subfamily 1 group H member 4; Farnesol receptor HRR-1; Farnesoid X-activated receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
417200 427.00
Check your price
  • NR5A1 Antibody (Monoclonal, N1665)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 434200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Steroid hormone receptor Ad4BP; Nuclear receptor subfamily 5 group A member 1; Fushi tarazu factor homolog 1; Adrenal 4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
434200 433.00
Check your price
  • ESRRA Antibody (Monoclonal, H5844)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 416600
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 3 group B member 1; Estrogen-related receptor alpha; Estrogen receptor-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
416600 433.00
Check your price
  • NuMA Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA316829
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:1000)
Alternate names:
SP-H antigen; Nuclear matrix protein-22
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316829 375.00
Check your price
  • Leishmania gp63 Antibody (Monoclonal, 96-126)
  • Host Mouse
  • Target Species Protozoa
  • Conjugate Unconjugated
  • Catalog # MA181830
  • Size 500 µL
Applications
ELISA (1:2000), Immunofluorescence (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181830 650.00
Check your price
  • TrkB Antibody (Monoclonal, 1.A.37)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191843
  • Size 25 µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tropomyosin-related kinase B; TrkB tyrosine kinase; Neurotrophic tyrosine kinase receptor type 2; GP145-TrkB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191843 375.00
Check your price
  • Survivin Antibody (Monoclonal, 2H5H2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517184
  • Size 100 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Apoptosis inhibitor survivin; Apoptosis inhibitor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517184 309.00
Check your price
  • Survivin Antibody (Monoclonal, 1H5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517035
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Apoptosis inhibitor survivin; Apoptosis inhibitor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517035 309.00
Check your price
  • XPG Antibody (Monoclonal, 8H7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126186
  • Size 100 µg
Applications
Immunoprecipitation (2 µg/mg of protein lysate), Western Blot (1:625-1:1250)
Alternate names:
Xeroderma pigmentosum group G-complementing protein; DNA excision repair protein ERCC-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126186 399.00
Check your price
  • PIK3R1 Antibody (Monoclonal, U5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA174183
  • Size 200 µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:100-1:1000)
Alternate names:
Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA174183 349.00
Check your price
  • Human ADAMTS1 Recombinant Protein
  • Host Insect cells
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP77543
  • Size 5 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP77543 446.00
Check your price
  • Tankyrase Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116550
  • Size 100 µL
Applications
Immunocytochemistry (1:250-1:500), Immunofluorescence (1:250-1:500), Western Blot (1:500-1:1000)
Alternate names:
Tankyrase I; TRF1-interacting ankyrin-related ADP-ribose polymerase; TNKS-1; Poly [ADP-ribose] polymerase 5A; ADP-ribosyltransferase diphtheria toxin-like 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116550 309.00
Check your price