1 - 3 of 3 products displayed
  • beta Lactamase Antibody (Monoclonal, 3E11.G3)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110713
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
blaCTX-M-14, beta-lactamase CTX-M-14, FLJ14902, G24, MRPL56
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110713 399.00
Check your price
  • beta Lactamase Antibody (Monoclonal, 8A5.A10)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA110712
  • Size 500 µg
Applications
ELISA (10-20 µg/ml), Western Blot (10 µg/ml)
Alternate names:
blaCTX-M-14, beta-lactamase CTX-M-14, FLJ14902, G24, MRPL56
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110712 399.00
Check your price
  • beta Lactamase Antibody (Monoclonal)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA120370
  • Size 500 µg
Applications
ELISA (10-20 µg/ml), Western Blot (5-15 µg/ml)
Alternate names:
blaCTX-M-14, beta-lactamase CTX-M-14, FLJ14902, G24, MRPL56
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120370 375.00
Check your price