1 - 30 of 30 products displayed
  • TCP-1 beta Monoclonal Antibody for Western Blot and IHC
  • Host Rat
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Non-human primate, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA190088
  • Size 50µg
Applications
Immunohistochemistry (Assay-dependent), Western Blot (1 µg/ml)
Alternate names:
CCT-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190088 375.00
Check your price
  • beta-2 Adaptin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11066
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11066 349.00
Check your price
  • IKK beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519916
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5 µg/ml)
Alternate names:
Nuclear factor NF-kappa-B inhibitor kinase beta; I-kappa-B kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519916 349.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # MA515739D488
  • Size 50µL
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D488 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA5157391MG
  • Size 1mg
Applications
ELISA (Assay dependent), Flow Cytometry (1-2 ug/test), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5157391MG 950.00
Check your price
  • TSH beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182909
  • Size 1mg
Applications
ELISA (Assay Dependent)
Alternate names:
Thyrotropin beta chain; Thyroid-stimulating hormone subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182909 325.00
Check your price
  • IkB beta Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540909
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (5 µg/ml)
Alternate names:
I-kappa-B-beta; Thyroid receptor-interacting protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540909 309.00
Check your price
  • beta Adducin Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Bovine, Dog
  • Conjugate Unconjugated
  • Catalog # PA543809
  • Size 100µl
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Erythrocyte adducin subunit beta; Adducin-63; Add97
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543809 309.00
Check your price
  • IFN beta Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520390
  • Size 100µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (5 ug/ml)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520390 349.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA515739D800
  • Size 50µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D800 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight® 680 conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 680
  • Catalog # MA515739D680
  • Size 50µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D680 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA515739BTIN
  • Size 50µL
Applications
Flow Cytometry (2 ug/test), Western Blot (1:500-1:1000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739BTIN 159.00
Check your price
  • beta Actin Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 550
  • Catalog # MA515739D550
  • Size 50µL
Applications
Immunocytochemistry (1:20-1:50), Immunofluorescence (1:20-1:50)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D550 159.00
Check your price
  • beta Actin Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 650
  • Catalog # MA515739D650
  • Size 50µl
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D650 159.00
Check your price
  • beta Actin Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate HRP
  • Catalog # MA515739HRP
  • Size 50µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739HRP 159.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516429
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (10 µg/ml), Western Blot (5-10 µg/mL)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516429 349.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA515739
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739 225.00
Check your price
  • beta Actin Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515452
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515452 349.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Leech, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516914
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (2-4 µg/ml)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516914 349.00
Check your price
  • beta Actin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121167
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121167 399.00
Check your price
  • Phospho-p70 S6 Kinase beta pSer423 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate
  • Catalog # PA535508
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
p70 ribosomal S6 kinase beta; Serine/threonine-protein kinase 14B; S6 kinase-related kinase; 70 kDa ribosomal protein S6 kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535508 Not Orderable Online
Check your price
  • HP1 beta Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528448
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p25beta; Modifier 1 protein; M31; Heterochromatin protein p25; Heterochromatin protein 1 homolog beta; HP1Hsbeta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528448 349.00
Check your price
  • HIF-1 beta Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and GS
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1515
  • Size 100µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:1,000), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:3000)
Alternate names:
Hypoxia-inducible factor 1-beta; Dioxin receptor, nuclear translocator; Class E basic helix-loop-helix protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1515 375.00
Check your price
  • PKA beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513794
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
cAMP-dependent protein kinase, beta-catalytic subunit; PKA C-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513794 349.00
Check your price
  • Phospho-PKC beta pSer661 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535445
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Protein kinase C beta type, PKC-B, PKC-beta, PRKCB, PKCB, PRKCB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535445 Not Orderable Online
Check your price
  • TSH beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183492
  • Size 1mg
Applications
ELISA (Assay Dependent)
Alternate names:
Thyrotropin beta chain; Thyroid-stimulating hormone subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183492 399.00
Check your price
  • TSH beta Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182908
  • Size 1mg
Applications
ELISA (Assay Dependent)
Alternate names:
Thyrotropin beta chain; Thyroid-stimulating hormone subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182908 325.00
Check your price
  • beta Tubulin Polyclonal Antibody for Western Blot, IHC (P), IP and FN
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121153
  • Size 500µL
Applications
Functional Assay (Assay-dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tubulin beta-5 chain; Tubulin beta-4 chain; Tubulin 5 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121153 390.00
Check your price
  • Estrogen Receptor beta Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1312
  • Size 50µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1-2 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Nuclear receptor subfamily 3 group A member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1312 349.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IF, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rodent, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519469
  • Size 100µg
Applications
ELISA (0.1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519469 349.00
Check your price